Skip to Content
Merck
All Photos(1)

Documents

46981

Supelco

Tylosin phosphate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H77NO17 · H3PO4
CAS Number:
Molecular Weight:
1014.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.[H][C@@]1(C[C@@](C)(O)[C@@H](O)[C@H](C)O1)O[C@@H]2[C@@H](C)O[C@@]([H])(O[C@@H]3[C@@H](C)[C@H](O)CC(=O)O[C@H](CC)[C@@H](CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC)\C=C(C)\C=C\C(=O)[C@H](C)C[C@@H]3CC=O)[C@H](O)[C@H]2N(C)C

InChI

1S/C46H77NO17.H3O4P/c1-13-33-30(22-58-45-42(57-12)41(56-11)37(52)26(5)60-45)18-23(2)14-15-31(49)24(3)19-29(16-17-48)39(25(4)32(50)20-34(51)62-33)64-44-38(53)36(47(9)10)40(27(6)61-44)63-35-21-46(8,55)43(54)28(7)59-35;1-5(2,3)4/h14-15,17-18,24-30,32-33,35-45,50,52-55H,13,16,19-22H2,1-12H3;(H3,1,2,3,4)/b15-14+,23-18+;/t24-,25+,26-,27-,28+,29+,30-,32-,33-,35+,36-,37-,38-,39-,40-,41-,42-,43+,44+,45-,46-;/m1./s1

InChI key

NBOODGNJLRRJNA-IAGPQMRQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahat Zaheer et al.
Frontiers in microbiology, 4, 133-133 (2013-06-12)
Macrolides are the first-line treatment against bovine respiratory disease (BRD), and are also used to treat infections in humans. The macrolide, tylosin phosphate, is often included in the diet of cattle as a preventative for liver abscesses in many regions
B Donev et al.
Veterinarno-meditsinski nauki, 20(5-6), 80-86 (1983-01-01)
Studied were both the acute and subchronic toxicity of technical tylosine phosphate (TP), produced at the Research Chemical and Pharmaceutical Institute, Sofia, used in combination with bentonite, known as Pharmasine TB. Pharmasine TB was practically untoxic for albino mice and
Yuranan Leknoi et al.
Journal of water and health, 15(2), 251-261 (2017-04-01)
We assessed the occurrence and specificity of bacteriophages of Bacteroides fragilis in swine farms for their potential application in microbial source tracking. A local B. fragilis host strain, SP25 (DSM29413), was isolated from a pooled swine feces sample taken from
[Comparative study of the distribution and excretion of tylosin of various origins].
M Kietzmann
DTW. Deutsche tierarztliche Wochenschrift, 92(4), 147-149 (1985-04-09)
Umut Gazi et al.
The Journal of biological chemistry, 286(10), 7822-7829 (2011-01-06)
The mannose receptor (MR) is an endocytic type I membrane molecule with a broad ligand specificity that is involved in both hemostasis and pathogen recognition. Membrane-anchored MR is cleaved by a metalloproteinase into functional soluble MR (sMR) composed of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service