Skip to Content
Merck
All Photos(1)

Key Documents

46382

Supelco

Florisil® PR Inorganic Sorbent

PR grade (analysis acc. to FDA), 60-100 mesh

Synonym(s):

Florisil®, Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

product name

Florisil® PR, PR grade (analysis acc. to FDA)

grade

PR grade (analysis acc. to FDA)

Quality Level

form

coarse powder

technique(s)

LPLC: suitable

surface area

289 m2/g

matrix

Magnesium silicate hydrate

matrix active group

magnesium silicate hydrate

particle size

149-250 μm
60-100 mesh

pore size

60 Å mean pore size

separation technique

hydrophilic interaction (HILIC)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Florisil, adsorbent for the separation of lipids, is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Application

Florisil was used:
  • as column packing material in column chromatography
  • as an adsorbent in gas chromatography with electron capture detector (GC/ECD), capillary column chromatography, adsorption column chromatography.
  • in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved gas chromatographic method for the determination of perfluorocarbon tracers in the atmosphere
Lagomarsino, Raymond J.
Journal of Chromatography A, 34, 405-412 (1996)
Determination of atmospheric perfluorocarbon background concentrations of fL/L range at the western coastal area of Korea
Kim, HyeKyeong, et al.
Atmospheric Environment, 1, 4-4 (2002)
R C Prados-Rosales et al.
Journal of chromatography. A, 993(1-2), 121-129 (2003-05-09)
A rapid analytical method for determination of organochlorine pesticide residues in sunflower seeds based on focused microwave-assisted Soxhlet extraction has been developed. The main factors affecting the extraction efficiency--namely microwave power, irradiation time, volume of extractant and number of cycles--were
The covalently bound flavin of Chromatium cytochrome c552. 1. Evidence for cysteine thiohemiacetal at the 8 alpha position.
W H Walker et al.
European journal of biochemistry, 48(2), 439-448 (1974-10-02)
P A de Paula Pereira et al.
Talanta, 49(2), 245-252 (2008-10-31)
A new sampling protocol was developed to determine methanol and ethanol in the gas phase, at low concentration levels, in urban atmospheres. The procedure involves collection of air samples (20.0-30.0 l) with three florisil cartridges connected in series, at a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service