Skip to Content
Merck
All Photos(1)

Documents

45495

Supelco

Fenvalerate

PESTANAL®, analytical standard

Synonym(s):

α-Cyano-3-phenoxybenzyl α-(4-chlorophenyl)­iso­valerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22ClNO3
CAS Number:
Molecular Weight:
419.90
Beilstein:
2025982
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C(C(=O)OC(C#N)c1cccc(Oc2ccccc2)c1)c3ccc(Cl)cc3

InChI

1S/C25H22ClNO3/c1-17(2)24(18-11-13-20(26)14-12-18)25(28)30-23(16-27)19-7-6-10-22(15-19)29-21-8-4-3-5-9-21/h3-15,17,23-24H,1-2H3

InChI key

NYPJDWWKZLNGGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Type II pyrethroid. Potent inhibitor of calcineurin (protein phosphatase 2B).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Terrence L Adelsbach et al.
Reviews of environmental contamination and toxicology, 176, 137-154 (2002-11-22)
Fenvalerate is listed under Class IV of the U.S. Food and Drug Administration (USFDA) Surveillance Index Classification, indicating a low hazard potential to humans from both exposure and toxicological standpoints; thus, minimal monitoring is required (Reed 1981; Eisler 1992). To
Xiaohua Gao et al.
Archives of toxicology, 86(9), 1443-1451 (2012-03-23)
Exposure to fenvalerate was demonstrated to be toxic to the male reproductive system. Our previous data revealed that intracellular calcium plays an important role in regulating the above toxicity, through actions on both T-type calcium channels and endoplasmic reticulum calcium
Nathalie C Stampfli et al.
Aquatic toxicology (Amsterdam, Netherlands), 127, 9-20 (2012-10-16)
Climate change models predict an increase in the frequency and intensity of extreme fluctuations in water level in aquatic habitats. Therefore, it is necessary to understand the combined effects of hydrological fluctuations and toxicants on aquatic biological communities. We investigated
Erin M Hitchner et al.
Journal of economic entomology, 105(1), 207-213 (2012-03-17)
Baseline toxicity levels to a novel semicarbazone insecticide, metaflumizone were established for 25 field populations of Colorado potato beetle, Leptinotarsa decemlineata Say (Coleoptera: Chrysomelidae),from North America. Excluding the susceptible laboratory strain, 50% lethal concentrations of metaflumizone ranged from 0.57 to
Lisbeth Schnug et al.
Environmental toxicology and chemistry, 32(4), 937-947 (2013-02-02)
The aim of the present study was to determine the toxicity of a mixture containing the biocides picoxystrobin, esfenvalerate, and triclosan to the reproduction and adult survival of two consecutive generations of Eisenia fetida (Savigny, 1826). Concentration addition and independent

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service