Skip to Content
Merck
All Photos(1)

Documents

45479

Supelco

Ethofumesate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O5S
CAS Number:
Molecular Weight:
286.34
Beilstein:
5759730
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC1Oc2ccc(OS(C)(=O)=O)cc2C1(C)C

InChI

1S/C13H18O5S/c1-5-16-12-13(2,3)10-8-9(18-19(4,14)15)6-7-11(10)17-12/h6-8,12H,5H2,1-4H3

InChI key

IRCMYGHHKLLGHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anne Chevillard et al.
Journal of hazardous materials, 205-206, 32-39 (2012-01-11)
The potential use of nanoclays for modulating transfer properties of active agents in bio-sourced polymers was explored. For this purpose, new pesticide formulations were designed by combining wheat gluten, ethofumesate (model pesticide) and three montmorillonites (MMT) using a bi-vis extrusion
D S Gardner et al.
Journal of agricultural and food chemistry, 49(6), 2894-2898 (2001-06-21)
The effect of turfgrass cover on the leaching and dissipation of ethofumesate and halofenozide was studied. Sampling cylinders (20 cm diam. x 30 cm long) were placed vertically in plots of creeping bentgrass (Agrostis palustris Huds.), tall fescue (Festuca arundinaceae
H B Runes et al.
Journal of agricultural and food chemistry, 47(8), 3252-3256 (1999-12-20)
Triadimefon, a fungicide, and ethofumesate, an herbicide, are commonly applied to turfgrass in the Pacific Northwest, resulting in foliar residues. A simple and rapid method was developed to determine triadimefon and ethofumesate concentrations from dislodgeable foliar residues on turfgrass. Turfgrass
Peng Xu et al.
Chemosphere, 112, 163-169 (2014-07-23)
Earthworms represent an important food source for many vertebrates and as a result, predators may encounter toxic effects via the food chain from consumption of contaminated worms. Therefore, including an assessment of xenobiotic to worms in risk assessment procedures is
Wentao Zhu et al.
Chirality, 19(8), 632-637 (2007-06-08)
The stereoselective toxicokinetics of ethofumesate enantiomers following a single intravenous (i.v.) administration at doses of 30 mg/kg were investigated in rabbits. Plasma concentrations of (+)- and (-)-ethofumesate were analyzed by a validated chiral HPLC method that involved extraction of plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service