Skip to Content
Merck
All Photos(1)

Key Documents

45453

Supelco

Dinoseb

PESTANAL®, analytical standard

Synonym(s):

2-sec-Butyl-4,6-dinitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O5
CAS Number:
Molecular Weight:
240.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)c1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C10H12N2O5/c1-3-6(2)8-4-7(11(14)15)5-9(10(8)13)12(16)17/h4-6,13H,3H2,1-2H3

InChI key

OWZPCEFYPSAJFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dinoseb may be used as a reference standard in the analysis of dinoseb residues in samples of raspberry extract using high performance liquid chromatography coupled with ultraviolet-visible light detector (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

306.9 °F - closed cup

Flash Point(C)

152.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatography method for the determination of dinoseb: application to the analysis of residues in raspberries
Szeto.YS and Price.MP
Journal of Agricultural and Food Chemistry, 39, 1614-1617 (1991)
Mark R Viant et al.
Aquatic toxicology (Amsterdam, Netherlands), 76(3-4), 329-342 (2005-11-18)
Changes in metabolism of Japanese medaka (Oryzias latipes) embryos exposed to dinoseb (2-sec-butyl-4,6-dinitrophenol), a substituted dinitrophenol herbicide, were determined by in vivo (31)P NMR, high-pressure liquid chromatography (HPLC)-UV, and (1)H NMR metabolomics. ATP and phosphocreatine (PCr) metabolism were characterized within
Mariko Matsumoto et al.
Environmental toxicology, 23(2), 169-183 (2008-01-25)
In a combined repeated dose toxicity study with reproduction/developmental toxicity screening test, Crj:CD(SD)IGS rats were dosed with dinoseb, 2-sec-butyl-4,6-dinitrophenol, by gavage at 0 (vehicle), 0.78, 2.33, or 7.0 mg/kg bw/day. Six males per group were dosed for a total of
Mariko Matsumoto et al.
Reproductive toxicology (Elmsford, N.Y.), 29(3), 292-297 (2010-02-06)
This study evaluated the prenatal developmental toxicity of the pesticide 2-sec-butyl-4,6-dinitrophenol (dinoseb). Pregnant rats were given dinoseb by gavage at 0, 8.0 or 10 mg/kg bw/day on days 6-15 of gestation, or in the diet at 0, 120 or 200
T Satapanajaru et al.
Journal of hazardous materials, 168(2-3), 930-937 (2009-04-07)
Dinoseb, a dinitroherbicide, was once commonly used in aerial crop dusting of agronomic crops in the western United States. Widespread use combined with improper disposal practices at rural air strips has contaminated numerous sites. Our objective was to determine if

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service