Skip to Content
Merck
All Photos(1)

Key Documents

45434

Supelco

Dichlon

PESTANAL®, analytical standard

Synonym(s):

2,3-Dichloro-1,4-naphthoquinone, Dichlon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4Cl2O2
CAS Number:
Molecular Weight:
227.04
Beilstein:
1073511
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

194-197 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)C(=O)c2ccccc2C1=O

InChI

1S/C10H4Cl2O2/c11-7-8(12)10(14)6-4-2-1-3-5(6)9(7)13/h1-4H

InChI key

SVPKNMBRVBMTLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlon is an agricultural fungicide of the quinone class, widely used against a variety of plant pathogenic fungi.

Application

Dichlon may be used as an analytical reference standard for the quantification of the analyte in food matrices using supercritical fluid extraction (SFE) and supercritical fluid chromatography (SFC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin-layer chromatography of some substituted naphthoquinones
Agricultural and Biological Chemistry, 30(9), 935-936 (1966)
Pesticide residues in canned foods, fruits, and vegetables: The application of supercritical fluid extraction and chromatographic techniques in the analysis
EL-Saeid.HM
TheScientificWorldJournal, 3(3), 1314-1326 (2003)
Chemistry and Biology of Water, Air and Soil: Environmental Aspects
Studies in Environmental Science, 53(3), 1314-1326 (1993)
S Ahmad et al.
Archives of environmental contamination and toxicology, 29(4), 442-448 (1995-11-01)
Southern armyworm, Spodoptera eridania, larvae were provided ad libitum 0.002-0.25% w/w dichlone, 2,3-dichloro-1,4-naphthoquinone (CNQ). Larval mortality occurred in a time-and-dose dependent manner, with an LC17 of 0.01% and an LC50 of 0.26% CNQ at day-5. Extracts of larvae fed control
Inhibition of liver glycolysis in rats by dietary dichlone (2,3-dichloro-1,4-naphthoquinone).
C A Pritsos et al.
Bulletin of environmental contamination and toxicology, 35(1), 23-28 (1985-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service