Skip to Content
Merck
All Photos(2)

Key Documents

45337

Supelco

Benfluralin

PESTANAL®, analytical standard

Synonym(s):

N-Butyl-2,6-dinitro-N-ethyl-4-trifluoromethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16F3N3O4
CAS Number:
Molecular Weight:
335.28
Beilstein:
2821329
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCN(CC)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

InChI

1S/C13H16F3N3O4/c1-3-5-6-17(4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3

InChI key

SMDHCQAYESWHAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Company ordered to halt sales of unapproved drugs, reimburse buyers.
FDA consumer, 38(5), 5-5 (2004-12-15)
Allergic contact dermatitis from the herbicides trifluralin and benefin.
M T Pentel et al.
Journal of the American Academy of Dermatology, 31(6), 1057-1058 (1994-12-01)
[Antiangiogenic and antitumor properties of cartilage].
D B Korman
Voprosy onkologii, 58(6), 717-726 (2012-01-01)
Sharks do get cancer: few surprises in cartilage research.
Joel B Finkelstein
Journal of the National Cancer Institute, 97(21), 1562-1563 (2005-11-03)
M Keseru et al.
Communications in agricultural and applied biological sciences, 69(4), 803-806 (2005-03-11)
The use of pesticides involves the risk of poisoning on wild animals. Teratological tests carried out on avian embryos provide useful data for environmental protection and facilitate the development of environment-friendly chemical plant protection techniques. A 30% dimethoate containing insecticide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service