Skip to Content
Merck
All Photos(2)

Key Documents

44527

Supelco

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

analytical standard

Synonym(s):

Caramel furanone, Sotolon, Sugar lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.491 (lit.)
n20/D 1.492

bp

184 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1OC(=O)C(O)=C1C

InChI

1S/C6H8O3/c1-3-4(2)9-6(8)5(3)7/h4,7H,1-2H3

InChI key

UNYNVICDCJHOPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one may be used as an analytical reference standard for quantification of the analyte in beer samples using gas chromatography (GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a stale-beer-like odorant in extracts of naturally aged beer
Callemien D, et al.
Journal of Agricultural and Food Chemistry, 54(4), 1409-1413 (2006)
Raquel M Callejón et al.
Journal of agricultural and food chemistry, 56(15), 6631-6639 (2008-07-12)
Two gas chromatography-olfactometry (GC-O) techniques were used to screen targeting compounds with an impact on the perceived quality of Sherry vinegar: detection frequency and aroma extract dilution analysis (AEDA). The GC-O study revealed the presence of 108 aromatic notes, of
Claudine Charpentier et al.
Antonie van Leeuwenhoek, 95(3), 263-273 (2009-02-18)
Fifty-four Saccharomyces cerevisiae strains were isolated from Jura "Vin Jaune" velum and characterized by conventional physiological and molecular tests including ITS RFLP and sequence analysis, karyotyping and inter delta typing. ITS RFLP and sequence revealed a specific group of related
Atsushi Taga et al.
Journal of oleo science, 61(1), 45-48 (2011-12-23)
A strongly aromatic compound, sotolon, was assessed by capillary zone electrophoresis within 9 min without specific pre-sample treatment. The calibration curve comprised a straight line with good linearity (R = 0.997) over a relatively wide range of 3.13 to 100
José Sousa Câmara et al.
Journal of agricultural and food chemistry, 52(22), 6765-6769 (2004-10-28)
The maturation of Madeira wines usually involves exposure to relatively high temperatures and humidity levels >70%, which affect the aroma and flavor composition and lead to the formation of the typical and characteristic bouquet of these wines. To estimate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service