Skip to Content
Merck
All Photos(1)

Key Documents

44064

Supelco

3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol

analytical standard

Synonym(s):

Ebanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O
CAS Number:
Molecular Weight:
208.34
Beilstein:
11329261
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(C(C)\C=C/C1C(C(=CC1)C)(C)C)C

InChI

1S/C14H24O/c1-10(12(3)15)6-8-13-9-7-11(2)14(13,4)5/h6-8,10,12-13,15H,9H2,1-5H3/b8-6-

InChI key

RNLHVODSMDJCBR-VURMDHGXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol, a derivative of α-campholenic aldehyde, is a synthetic alternative for sandalwood oil − a key perfume ingredient.

Application

3-Methyl-5-(2,2,3-trimethyl-3-cyclopenten-1-yl)-4-penten-2-ol may be used as an analytical standard for the determination of the analyte in sandalwood oil (Santalum album L.), cosmetics and personal care products, and biological samples of the animals exposed to the sandalwood odorants by gas chromatography (GC) based techniques.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

226.4 °F

Flash Point(C)

108 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Four-stage (low-) flow modulation comprehensive gas chromatography? quadrupole mass spectrometry for the determination of recently-highlighted cosmetic allergens
Tranchida PQ, et al.
Journal of Chromatography A, 1439(1), 144-151 (2016)
Authenticity of essential oils
Do TKT, et al.
TrAC, Trends in Analytical Chemistry, 66(6), 146-157 (2015)
Olfactory receptor neuron profiling using sandalwood odorants
Bieri S, et al.
Chemical Senses, 29(6), 483-487 (2004)
Danilo Sciarrone et al.
Journal of chromatography. A, 1218(1), 137-142 (2010-11-30)
The production and trade of Indian sandalwood oil is strictly regulated, due to the impoverishment of the plantations; for such a reason, Australian sandalwood oil has been evaluated as a possible substitute of the Indian type. International directives report, for
The survey of analytical methods for sample preparation and analysis of fragrances in cosmetics and personal care products
Abedi G, et al.
TrAC, Trends in Analytical Chemistry, 102(1), 41-59 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service