Skip to Content
Merck
All Photos(1)

Key Documents

43866

Supelco

Salidroside

analytical standard

Synonym(s):

2-(4-Hydroxyphenyl)ethyl β−D-glucopyranoside, p-Hydroxyphenethyl glucopyranoside, Rhodioloside, Rhodosin, Tyrosol a-(β-D-glucopyranoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O7
CAS Number:
Molecular Weight:
300.30
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -29±2°, c = 0.5 in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@@H](OCCc2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H20O7/c15-7-10-11(17)12(18)13(19)14(21-10)20-6-5-8-1-3-9(16)4-2-8/h1-4,10-19H,5-7H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

ILRCGYURZSFMEG-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Salidroside is one of the major phenolic glycoside extracted from Rhodiola rosea L([1], which can show a broad-spectrum of pharmacological properties like anti-aging, anti-oxidant, anti-cancer and anti-inflammation.[2][1][3]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Salidroside, the 8-O-β-D-glucoside of tyrosol, is the main bioactive component of Rhodiola species and is found mainly in the plant roots. It is biosynthesised in the tyrosine metabolism, with glucosylation of tyrosol being the final step[4]. It has been reported to be neuroprotective[2] [5][3] and one of the compounds responsible for the antidepressant and anxiolytic actions of Rhodiola rosea[6][7].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wieland Peschel et al.
Phytochemistry, 86, 92-102 (2012-11-10)
The demand for plant material of Rhodiola rosea L. (Crassulaceae) for medicinal use has increased recently, amid concerns about its quality and sustainability. We have analysed the content of phenylpropanoids (total rosavins) and salidroside in liquid extracts from 3-year old
Ze-qiang Qu et al.
PloS one, 7(1), e29641-e29641 (2012-01-12)
Previously we have demonstrated that a Rhodiola crenulata extract (RCE), containing a potent antioxidant salidroside, promotes neurogenesis in the hippocampus of depressive rats. The current study was designed to further investigate the protective effect of the RCE on neurogenesis in
Jin-Kang Zhang et al.
PloS one, 8(2), e57251-e57251 (2013-02-26)
Oxidative stress is a pivotal pathogenic factor for bone loss in mouse model. Salidroside, a phenylpropanoid glycoside extracted from Rhodiola rosea L, exhibits potent antioxidative effects. In the present study, we used an in vitro oxidative stress model induced by
Ken Yu-Zhong Zheng et al.
European journal of pharmacology, 679(1-3), 34-39 (2012-02-09)
Rhodiolae Crenulatae Radix et Rhizoma (Rhodiola), the root and rhizome of Rhodiola crenulata (Hook. f. et Thoms.) H. Ohba, has been used as a traditional Chinese medicine (TCM) to increase the body resistance to mountain sickness in preventing hypoxia; however
A Panossian et al.
Phytotherapy research : PTR, 19(10), 819-838 (2005-11-02)
Plant adaptogens are compounds that increase the ability of an organism to adapt to environmental factors and to avoid damage from such factors. The beneficial effects of multi-dose administration of adaptogens are mainly associated with the hypothalamic-pituitary-adrenal (HPA) axis, a

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service