Skip to Content
Merck
All Photos(1)

Key Documents

43412

Supelco

(+)-Catechin

analytical standard

Synonym(s):

(+)-Cyanidanol, D-Catechin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1

InChI key

PFTAWBLQPZVEMU-DZGCQCFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Catechin is the enantiomer of the polyphenolic flavonoid, catechin. Known for its antioxidative nature, it is present in various herbs, vegetables, fruits, green teas, chocolates, and red wines.[1][2]

Application

This analytical standard can also be used as follows:
  • Method development for the multi-determination of catechin and its diastereomers in the leaf extracts and infusions obtained from five Byrsonima species using high-performance liquid chromatography, combined with photodiode array (PAD) and circular dichroism (CD) detectors
  • Determination of catechin in commercial green and black tea samples using magnetic solid phase extraction (MSPE) based on a novel adsorbent— O-carboxymethyl-chitosan-g- acrylic acid sodium salt (O-CMCs-g-AAS) copolymer deposited on magnetic graphene oxide (MGO), for sample pre-treatment and gas chromatography-mass spectrometry (GC-MS)
  • Multi-residue analysis of eight catechins and four theaflavins in three different commercial tea infusions (green, black, and oolong) by ultra high-performance liquid chromatography in combination with triple-quadrupole tandem mass spectrometry (UHPLC-MS-MS)
  • Simultaneous detection and quantification of four phenolic compounds— catechin, caffeic acid, rutin, and trans-cinnamic acid, using ultrasonic-assisted extraction and HPLC coupled with diode array detection from Physalis angulata L. plant material samples
  • Multi-residue determination of catechins, caffeine, and polyphenols, from 45 fresh Jukro tea leaf samples, collected at 40, 60, and 90-day intervals, by an HPLC-based method combined with PDA detection

Packaging

Bottomless glass bottle. Content inside is inserted in a fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meran Keshawa Ediriweera et al.
Medicines (Basel, Switzerland), 4(3) (2017-09-21)
Obesity is considered as one of the risk factors for breast cancer. Leptin has been found to be involved in breast cancer progression. Therefore, novel approaches to antagonize biological effects of leptin are much needed. The objective of this study
Catechin contents of foods commonly consumed in The Netherlands. 1. Fruits, vegetables, staple foods, and processed foods
Arts.W.C.I, et al.
Journal of Agricultural and Food Chemistry, 48(5), 1746-1751 (2000)
Catechin contents of foods commonly consumed in The Netherlands. 1. Fruits, vegetables, staple foods, and processed foods
Arts.W.C.I, et al.
Journal of Agricultural and Food Chemistry, 48, 1746-1751 (2000)
(+)-Catechin in human plasma after ingestion of a single serving of reconstituted red wine
Bell RCJ, et al
American Journal of Clinical Nutrition, 71, 103-108 (2000)
Andrej Živković et al.
Foods (Basel, Switzerland), 10(4) (2021-05-01)
The aim was to investigate the effects of the cold dehulling of buckwheat seeds on their germination, total phenolic content (TPC), antioxidant activity (AA) and phenolics composition. Cold dehulling had no negative effects on germination rate and resulted in faster

Articles

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service