Skip to Content
Merck
All Photos(1)

Key Documents

41700

Sigma-Aldrich

4,N,N-Trimethylaniline

purum, ≥98.0% (GC)

Synonym(s):

4-Dimethylaminotoluene, N,N-Dimethyl-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(CH3)2
CAS Number:
Molecular Weight:
135.21
Beilstein:
774409
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

grade

purum

Assay

≥98.0% (GC)

form

liquid

expl. lim.

7 %

refractive index

n20/D 1.546 (lit.)
n20/D 1.547

bp

211 °C (lit.)
90-92 °C/10 mmHg (lit.)

density

0.937 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)c1ccc(C)cc1

InChI

1S/C9H13N/c1-8-4-6-9(7-5-8)10(2)3/h4-7H,1-3H3

InChI key

GYVGXEWAOAAJEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,N,N-Trimethylaniline is a N-methyl-N-alkylaniline. Its reaction with vinyl ether catalyzed by CuCl2 has been reported to afford tetrahydroquinolines. Its radical cation undergoes reaction with the anthracene radical anion and generation of electrogenerated chemiluminescence (ECL) has been observed.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Reproductive organs

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob B Ketter et al.
Journal of the American Chemical Society, 126(32), 10183-10189 (2004-08-12)
Electrogenerated chemiluminescence (ECL) arising from the reaction of radical ions has previously be shown to arise from a variety of states including excited singlets, triplets, excimers, and exciplexes. In this work we describe two systems that form emissive states in
Zoe M Wright et al.
Advanced healthcare materials, 10(2), e2001189-e2001189 (2020-12-17)
Graphene oxide and functionalized graphenic materials (FGMs) have promise as platforms for imparting programmable bioactivity to poly(methyl methacrylate) (PMMA)-based bone cement. To date, however, graphenic fillers have only been feasible in PMMA cements at extremely low loadings, limiting the bioactive
Anuradha Prakki et al.
Dental materials : official publication of the Academy of Dental Materials, 25(1), 26-32 (2008-09-02)
The purpose of this study was to evaluate the effect of two additives, propionaldehyde/aldehyde or 2,3-butanedione/diketone, on mechanical properties of Bis-GMA-based composites containing TEGDMA, propoxylated Bis-GMA (CH(3)Bis-GMA) or propoxylated fluorinated Bis-GMA (CF(3)Bis-GMA). Three control composites, Bis-GMA/diluent monomer (25/75 mol%), and
Chiemi Hirabayashi et al.
Dental materials journal, 21(4), 314-321 (2003-03-01)
Polymerization characteristics of poly (methyl methacrylate)(PMMA)/(methyl methacrylate) (MMA) resin initiated by tributylborane (TBB) were compared with those by benzoyl peroxide (BPO)/N,N-dimethyl-p-toluidine and camphorquinone (CQ)/N,N-dimethylaminoethyl methacrylate from the aspects of temporal changes of residual MMA and molecular weight up to 4
Kyle Winter et al.
Biomaterials, 26(26), 5321-5329 (2005-04-09)
Previous investigations have found that visible-light (VL)-irradiated camphorquinone (CQ), in the presence of a tertiary amine (e.g., N,N-dimethyl-p-toluidine, DMT), generates reactive oxygen species and causes oxidative DNA damage in vitro. In this study, oxidative DNA damage produced by VL-irradiated CQ/DMT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service