Skip to Content
Merck
All Photos(1)

Key Documents

37901

Supelco

Isocarbophos

PESTANAL®, analytical standard

Synonym(s):

O-Methyl O-[2-(isopropoxycarbonyl)phenyl] phosphoramidothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16NO4PS
CAS Number:
Molecular Weight:
289.29
Beilstein:
2946521
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(N)(=S)Oc1ccccc1C(=O)OC(C)C

InChI

1S/C11H16NO4PS/c1-8(2)15-11(13)9-6-4-5-7-10(9)16-17(12,18)14-3/h4-8H,1-3H3,(H2,12,18)

InChI key

YFVOXLJXJBQDEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Jia et al.
Ecotoxicology and environmental safety, 163, 1-6 (2018-07-22)
Although isocarbophos is a widely used insecticide, its toxicity to aquatic organisms has not been well characterized. In this study, zebrafish were exposed to isocarbophos at concentrations of 50 µg L-1 and 200 µg L-1 to assess its bioaccumulation, metabolic disruption, and oxidative stress.
Xiaotong Li et al.
Talanta, 188, 232-237 (2018-07-22)
A novel label-free and enzyme-free detection strategy has been developed for the fluorescent detection of isocarbophos (ICP) using multi-walled carbon nanotubes (MWCNTs) and G-quadruplex as the signal transducers. In this work, the split ICP aptamer were attached to a G-quadruplex
Huixiang Ouyang et al.
Frontiers in chemistry, 8, 673-673 (2020-09-15)
Fullerol (C60OH) has been shown to catalyze the trisodium citrate (TSC)-silver nitrate reaction to generate Ag nanoparticles (AgNPs). These AgNPs exhibit significant nanoplasmic surface-enhanced Raman scattering (SERS), resonance Rayleigh scattering (RRS), and absorption (Abs). When an aptamer (Apt) adsorbs on
Jian Zhang et al.
Biosensors & bioelectronics, 150, 111879-111879 (2019-11-27)
With increasing industrialization of food production, residues of organophosphorus pesticides (OPs) are more frequently found in the environment including rivers, lakes and soils. Extended exposure to OPs, even at a level below 1 nM, may lead to liver and central nervous
Xiaoxia Li et al.
Sensors (Basel, Switzerland), 18(12) (2018-12-20)
A simple and dual modal (colorimetric and fluorescent) sensor for organophosphate pesticides with high sensitivity and selectivity using adenosine triphosphate (ATP)- and rhodamine B-modified gold nanoparticles (RB-AuNPs), was successfully fabricated. This detection for ethoprophos afforded colorimetric and fluorescence imaging changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service