Skip to Content
Merck
All Photos(1)

Key Documents

37897

Supelco

Bensulfuron-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O7S
CAS Number:
Molecular Weight:
410.40
Beilstein:
7447488
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤1% water (Karl Fischer)

mp

179-184 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccccc1CS(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C16H18N4O7S/c1-25-12-8-13(26-2)18-15(17-12)19-16(22)20-28(23,24)9-10-6-4-5-7-11(10)14(21)27-3/h4-8H,9H2,1-3H3,(H2,17,18,19,20,22)

InChI key

XMQFTWRPUQYINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Sensitive, handle under argon.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chui-Hua Kong et al.
Pest management science, 64(3), 276-282 (2008-01-04)
A number of techniques, including cultural management, allelopathy and bioherbicide, have been considered as alternatives for synthetic herbicides, but successful weed control will require the careful integration of these multiple techniques. This study was conducted to assess the use of
Xiao-Yan Lin et al.
Ecotoxicology (London, England), 21(5), 1281-1290 (2012-03-21)
Bensulfuron-methyl (BSM) is a new kind of sulfonylurea herbicide widely used to control broad-leaf weeds in rice paddies. The aim of this work was to study BSM biodegradation in paddy soils with BSM-degrading bacteria Bacillus megaterium L1 and Brevibacterium sp.
Xiaoyan Lin et al.
Ecotoxicology and environmental safety, 72(7), 1899-1904 (2009-05-30)
Oxidative stress enzymes, superoxide dismutase (SOD), catalase (CAT), and ATPase, from two Gram-positive bacteria and one Gram-negative bacterium, respectively, were tested for response to the oxidative stress caused by bensulfuron-methyl (BSM). Native PAGE was used to detect the SOD isoenzyme
Wei Zhang et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 18(3), 613-619 (2007-06-08)
By the methods of batch equilibration and leaching, this paper studied the effects of solution pH and simulated acid rain on the behavior of bensulfuron-methyl and metsulfuron-methyl in soil. The results showed that the adsorption isotherms of these two herbicides
Kaijie Tang et al.
Analytica chimica acta, 614(1), 112-118 (2008-04-15)
A pre-treatment methodology based on the molecularly imprinted solid phase extraction (MI-SPE) procedure was developed for the determination of bensulfuron-methyl (BSM), tribenuron-methyl (TBM), metsulfuron-methyl (MSM) and nicosulfuron (NS) in soybean samples. A molecular imprinted polymer (MIP) was prepared by precipitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service