Skip to Content
Merck
All Photos(1)

Key Documents

36138

Supelco

Isoxaben

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O4
CAS Number:
Molecular Weight:
332.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

175-179 °C

application(s)

agriculture
environmental

format

neat

SMILES string

CCC(C)(CC)c1cc(NC(=O)c2c(OC)cccc2OC)on1

InChI

1S/C18H24N2O4/c1-6-18(3,7-2)14-11-15(24-20-14)19-17(21)16-12(22-4)9-8-10-13(16)23-5/h8-11H,6-7H2,1-5H3,(H,19,21)

InChI key

PMHURSZHKKJGBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoxaben is a pre-emergence, broad leaf herbicide, which can be used on turf grass, small grains and ornamental type of plants in order to basically inhibit the synthesis of cellulose.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isaac Shim et al.
Frontiers in plant science, 9, 1152-1152 (2018-09-11)
The cellulose synthase (CESA) proteins in Arabidopsis play an essential role in the production of cellulose in the cell walls. Herbicides such as isoxaben and flupoxam specifically target this production process and are prominent cellulose biosynthesis inhibitors (CBIs). Forward genetic
Novel cell wall architecture of isoxaben?habituated Arabidopsis suspension?cultured cells: global transcript profiling and cellular analysis
Manfield.WL
The Plant Journal, 40, 260-275 (2004)
Alexander R Paredez et al.
Science (New York, N.Y.), 312(5779), 1491-1495 (2006-04-22)
Expression of a functional yellow fluorescent protein fusion to cellulose synthase (CESA) in transgenic Arabidopsis plants allowed the process of cellulose deposition to be visualized in living cells. Spinning disk confocal microscopy revealed that CESA complexes in the plasma membrane
Ana Caño-Delgado et al.
The Plant journal : for cell and molecular biology, 34(3), 351-362 (2003-04-26)
The cell wall determines the shape of plant cells and is also the primary interface for pathogen interactions. The structure of the cell wall can be modified in response to developmental and environmental cues, for example to strengthen the wall
Jonatan U Fangel et al.
Plant science : an international journal of experimental plant biology, 180(3), 470-479 (2011-03-23)
Plant cell wall polysaccharides are amongst the most complex, heterogeneous and abundant bio-molecules on earth. This makes the biosynthetic enzymes, namely the glycosyltransferases and polysaccharide synthases, important research targets in plant science and biotechnology. As an initial step to characterize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service