Skip to Content
Merck
All Photos(1)

Documents

35995

Supelco

Eugenol

PESTANAL®, analytical standard

Synonym(s):

2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1366759
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

99.6% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)

bp

254 °C (lit.)

mp

−12-−10 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(CC=C)ccc1O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

Eugenol is a naturally occurring flavor extract isolated from the plants, including kernels of Myristica fragrans Houtt, bark of the genus Cinnamonum, roots of Valeriana officinalis, etc. It finds applications as a substrate for vanillin production via enzymatic transformation. Eugenol is released from clove-bud-containing cigarettes, and reportedly exhibits analgesic property, which helps to reduce the pain in the lungs and throat by freezing the pain receptors in the mouth and throat.
Eugenol is a phenolic compound and a major constituent of clove oil, which can find applications as an anti H. pylori agents and anti-bacterial agents. It can also act like a fungicidal agent, which can completely inhibit the growth of Saccharomyces cerevisiae, a species of yeast.

Application

Eugenol may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Bioconversion broth using reversed-phase high-performance liquid chromatography with UV detection (RP-HPLC-UV).
  • Mainstream cigarette smoke using gas chromatography/mass spectrometry (GC/MS).
  • Plant extracts using high-performance thin-layer chromatography (HPTLC) and gas chromatography coupled to mass spectrometry (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: angelica cinnamomum

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of eugenol, anethole, and coumarin in the mainstream cigarette smoke of Indonesian clove cigarettes
Polzin MG, et al.
Food And Chemical Toxicology, 45(10), 1948-1953 (2007)
HPTLC and GC-MS for separation and identification of eugenol in plants
Rusu LM, et al.
J. Planar Chromatogr., 20(2), 139-140 (2007)
Antimicrobial activities of Eugenol and Cinnamaldehyde against the human gastric pathogen Helicobacter pylori
Ali MS, et al.
Annals of Clinical Microbiology and Antimicrobials, 4, 20-20 (2005)
Surface alteration of Saccharomyces cerevisiae induced by thymol and eugenol
Bennis.S, et al.
Letters in Applied Microbiology, 38, 454-458 (2004)
Determination of vanillin, eugenol and isoeugenol by RP-HPLC.
Li-H-Y, et al.
Chromatographia, 60(11-12), 709-713 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service