Skip to Content
Merck
All Photos(2)

Documents

35599

Supelco

PCB No 52

analytical standard

Synonym(s):

2,2′,5,5′-Tetrachlorobiphenyl, 2,2′,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl4
CAS Number:
Molecular Weight:
291.99
Beilstein:
2053828
Ballschmiter Number:
52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(Cl)c(c1)-c2cc(Cl)ccc2Cl

InChI

1S/C12H6Cl4/c13-7-1-3-11(15)9(5-7)10-6-8(14)2-4-12(10)16/h1-6H

InChI key

HCWZEPKLWVAEOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls (PCBs) are groups of chemical compounds which are persistent in the environment and are considered highly toxic to humans and animals. PCB No 52 may be used for environmental analysis and precise quality control of food and feed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Schäfer et al.
Archives of biochemistry and biophysics, 488(1), 60-68 (2009-07-01)
There are 75 full length cytochrome P450 (CYP) genes known in the genome of the nematode Caenorhabditis elegans. The individual biological functions of the vast majority are mostly as yet unknown. Here the impact of cytochrome P450 isoforms on the
Bayram Yilmaz et al.
Environmental toxicology, 27(3), 185-191 (2011-02-24)
Cultured porcine endothelial cells were used to determine the effects of several congeners of polychlorinated biphenyls (PCBs) on cell viability and changes induced by these congeners on levels of intracellular calcium and reactive oxygen species (ROS). Cultured endothelial cells were
Ze-Yu Yang et al.
Chemosphere, 72(10), 1435-1440 (2008-07-04)
To verify a theoretical mass balance and multiple compartment partitioning model developed to predict freely dissolved concentrations (FDCs) of hydrophobic organic chemicals (HOCs) using negligible depletion-solid phase microextraction (nd-SPME), a series of sediment slurry experiments were performed using disposable poly(dimethyl)siloxane
Suleyman Sandal et al.
Immunopharmacology and immunotoxicology, 27(4), 601-613 (2006-01-27)
Polychlorinated biphenyls (PCBs) are industrial chemicals that have been released into the environment, resulting in widespread and persistent contamination. PCBs exist as 209 different congeners depending on the chlorine substitution on the biphenyl rings, and the physical properties and toxic
R J Jandacek et al.
Environment international, 36(8), 880-883 (2009-07-21)
Mice were gavaged with either (14)C-labeled 2,2'5,5' tetrachlorobiphenyl; 3,3',4,4' tetrachlorobiphenyl; or perfluorooctanoic acid. Absorption of these compounds was determined by assay of feces collected for 48 h after the gavage. Part of the animals received test diets containing olestra during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service