Skip to Content
Merck
All Photos(1)

Key Documents

35029

Supelco

Triflumuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClF3N2O3
CAS Number:
Molecular Weight:
358.70
Beilstein:
2776684
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)(F)Oc1ccc(NC(=O)NC(=O)c2ccccc2Cl)cc1

InChI

1S/C15H10ClF3N2O3/c16-12-4-2-1-3-11(12)13(22)21-14(23)20-9-5-7-10(8-6-9)24-15(17,18)19/h1-8H,(H2,20,21,22,23)

InChI key

XAIPTRIXGHTTNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triflumuron is a non-sysmetic insect growth regulator or insecticide, which can inhibit chitin synthesis. Its can act against moulting the larvae and also cause infertility of the eggs.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stewart Lowden et al.
Veterinary parasitology, 148(3-4), 295-300 (2007-08-11)
The horse biting louse (Werneckiella equi) is a common global equine ectoparasite. To our knowledge, benzoyl(phenyl)urea insecticides (triflumuron, diflubenzuron) commonly used as sheep lousicides, have not been evaluated for efficacy against W. equi. The aim of this study was to
C P Batra et al.
Journal of vector borne diseases, 42(3), 109-116 (2005-11-22)
An environmental friendly formulation Starycide 480 SC (Triflumuron-OMS-2015 ), a new insect growth regulator with chitin synthesis inhibitor type mode of action was evaluated against mosquito larvae in laboratory and small-scale field trials carried out in and around Delhi. The
T S Waghorn et al.
New Zealand veterinary journal, 61(5), 274-280 (2013-02-28)
To evaluate the in vitro efficacy of triflumuron and dicyclanil against field strains of Lucilia sericata and L. cuprina in New Zealand to establish whether anecdotal reports of declining protection periods afforded by dips containing these active ingredients had validity.
P J James et al.
Medical and veterinary entomology, 22(4), 326-330 (2009-01-06)
Low-volume, backline applications with the benzoylphenyl urea insecticides triflumuron and diflubenzuron represent in excess of 70% of treatments for the control of sheep lice, Bovicola ovis (Schrank) (Phthiraptera: Trichodectidae), in Australia. Reports of reduced effectiveness from 2003 and subsequent controlled
Autosterilization of the house fly, Musca domestica, using the chitin synthesis inhibitor triflumuron on sugar-baited targets.
J Howard et al.
Medical and veterinary entomology, 10(1), 97-100 (1996-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service