Skip to Content
Merck
All Photos(1)

Key Documents

34198

Supelco

Ractopamine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

4-{3-[2-Hydroxy-2-(4-hydroxyphenyl)ethyl]aminobutylphenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO3 · HCl
CAS Number:
Molecular Weight:
337.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC1=CC=C(C(O)CNC(C)CCC2=CC=C(O)C=C2)C=C1.Cl

InChI

1S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22H,2-3,12H2,1H3;1H

InChI key

JHGSLSLUFMZUMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ractopamine hydrochloride is a phenethanolamine β-adrenergic agonist (β-AA) that improves growth performance and affects carcass characteristics in livestock, through increased protein synthesis. Ractopamine hydrochloride is FDA approved animal feed additive used in some third countries like the USA, Canada, Japan, and Mexico for the growth promotion in fattening of pigs and cattle. In the USA, Ractopamine hydrochloride is authorized for use in pigs and cattle at a feed concentration of 5-20 mg kg-1 for finishing pigs and 10-30 mg kg-1 for cattle in confinement.

Application

The analytical standard can be used to:
  • Evaluate the comparative effects of ractopamine hydrochloride and zilpaterol hydrochloride on growth performance, carcass traits, and longissimus tenderness of finishing beef steers
  • Determine the effects of ractopamine hydrochloride on growth performance, carcass characteristics, physiological response to handling, and mobility during shipment for slaughter
  • Evaluate the effect of ractopamine hydrochloride on growth performance, carcass cutting yield, and processing characteristics of boneless hams and bellies from finishing pigs
  • Study the effects of ractopamine hydrochloride and zilpaterol hydrochloride on growth, diet digestibility, intake, and carcass characteristics of feedlot lambs
  • Evaluate the effects of ractopamine hydrochloride on growth performance, feed intake patterns, and acid-base balance in feedlot cattle

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Hagenmaier et al.
Journal of animal science, 95(5), 1977-1992 (2017-07-21)
Feedlot cattle ( = 128; BW = 549 ± 60 kg) were used to evaluate the effects of ractopamine hydrochloride (RAC) on growth performance, physiological response to handling, and mobility during shipment for slaughter in a study utilizing a split-plot
C S Abney et al.
Journal of animal science, 85(11), 3090-3098 (2007-07-05)
Two experiments evaluated effects of ractopamine hydrochloride (RAC) on performance, intake patterns, and acid-base balance of feedlot cattle. In Exp. 1, 360 crossbred steers (Brangus, British, and British x Continental breeding; initial BW = 545 kg) were used in a
L E Watkins et al.
Journal of animal science, 68(11), 3588-3595 (1990-11-01)
Six trials involving 888 pigs (Study 1) and three trials involving 360 pigs (Study 2) were conducted at various geographical locations in the U.S. and Canada to evaluate the effect of ractopamine hydrochloride on the performance and carcass characteristics of
Effect of ractopamine hydrochloride and zilpaterol hydrochloride on growth, diet digestibility, intake and carcass characteristics of feedlot lambs
Lopez-Carlos MA, et al.
Livestock Science, 23-30 null
Safety evaluation of ractopamine
European Food Safety Authority
EFSA Journal, 1041 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service