Skip to Content
Merck
All Photos(2)

Key Documents

33739

Supelco

Pyrethrum extract

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\C1[C@@H](C(=O)O[C@H]2CC(=O)C(C\C=C\C=C)=C2C)C1(C)C.COC(=O)\C(C)=C\C3[C@@H](C(=O)OC4CC(=O)C(C\C=C\C=C)=C4C)C3(C)C

InChI

1S/C22H28O5.C21H28O3/c1-7-8-9-10-15-14(3)18(12-17(15)23)27-21(25)19-16(22(19,4)5)11-13(2)20(24)26-6;1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h7-9,11,16,18-19H,1,10,12H2,2-6H3;7-9,11,16,18-19H,1,10,12H2,2-6H3/b9-8+,13-11+;9-8+/t16?,18?,19-;16?,18-,19-/m00/s1

InChI key

VXSIXFKKSNGRRO-YWUDCVDHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrethrum extract is an extract of pyrethrum flowers in petroleum oil with a definite pyrethrin content.[1] It instantly brings about a knock down action in insects, thereby acting as an effective insecticide.[2]

Application

Pyrethrum extract may be used to obtain natural pyrethrins, separated using high resolution gas chromatographic analysis(HRGC)[3], which can serve as active ingredients in insecticidal formulations for indoor use[3], agricultural and also in industrial pest management.[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of the insecticidal pyrethrin esters by capillary electrochromatography
III Henry.WC, et al.
Journal of Chromatography A, 905, 319-327 (2001)
Textbook of Pharmacognosy and Phytochemistry (2009)
Pyrethroids as household insecticides: analysis, indoor exposure and persistence
Class.JT and Kin J
Fresenius Journal of Analytical Chemistry, 340, 446-453 (1991)
The beet leafhopper and its control on beets grown for seed in Arizona and New Mexico, Volumes 851-875 (1943)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service