Skip to Content
Merck
All Photos(1)

Key Documents

33461

Supelco

N-(1-Naphthyl)ethylenediamine dihydrochloride

ACS reagent, ≥98%

Synonym(s):

2-(1-Naphthylamino)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
Beilstein:
3707471
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥98% (calc with ref. to anhyd. subst.)
≥98%

form

powder

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤5% water (Karl Fischer)

mp

194-198 °C (dec.) (lit.)

suitability

complies for identity

SMILES string

Cl.Cl.NCCNc1cccc2ccccc12

InChI

1S/C12H14N2.2ClH/c13-8-9-14-12-7-3-5-10-4-1-2-6-11(10)12;;/h1-7,14H,8-9,13H2;2*1H

InChI key

MZNYWPRCVDMOJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(1-Naphthyl)ethylenediamine dihydrochloride dissolved in sulfuric acid-methanol may be used as a reagent for the determination of sugar on thin layer plates.[1] It may also be used for the determination of sub-micromolar concentrations of nitrite ion after diazotisation with sulphanilic acid at low pH, by differential-pulse polarography.[2] It may also be used as the coupling agent for the spectrophotometric determination of aniline by the diazotization-coupling method.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of aniline by the diazotization-coupling method with N-(1-naphthyl) ethylenediamine as the coupling agent.
Norwitz G and Keliher PN.
Analytical Chemistry, 53 (8), 1238-1240 (1981)
Determination of nitrite ion by differential-pulse polarography using N-(1-naphthyl) ethylenediamine.
Sabharwal S.
Analyst, 115 (10), 1305-1307 (1990)
N-(1-Naphthyl) ethylenediamine dihydrochloride as a new reagent for nanomole quantification of sugars on thin-layer plates by a mathematical calibration process.
Bounias M.
Analytical Chemistry, 106 (2), 291-295 (1980)
Shigenori Nakano et al.
Talanta, 81(3), 786-791 (2010-03-20)
A novel flow-injection spectrophotometry has been developed for the determination of molybdenum(VI) at nanograms per milliliter levels. The method is based on the catalytic effect of molybdenum(VI) on the bromate oxidative coupling of p-hydrazinobenzenesulfonic acid with N-(1-naphthyl)ethylenediamine to form an
Improved dye procedure for determining urea concentration by using o-phthalaldehyde and naphthylethylenediamine.
N T Lequang et al.
Clinical chemistry, 33(1), 192-192 (1987-01-01)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service