Skip to Content
Merck
All Photos(1)

Key Documents

33374

Supelco

Nitrofen

PESTANAL®, analytical standard

Synonym(s):

2,4-Dichlorophenyl 4-nitrophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl2NO3
CAS Number:
Molecular Weight:
284.09
Beilstein:
1887356
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

69-72 °C

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[O-][N+](=O)c1ccc(Oc2ccc(Cl)cc2Cl)cc1

InChI

1S/C12H7Cl2NO3/c13-8-1-6-12(11(14)7-8)18-10-4-2-9(3-5-10)15(16)17/h1-7H

InChI key

XITQUSLLOSKDTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrofen can be used as a herbicide in order to control weeds in various crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

National study of chemical residues in fish Volume II null
Ya-Ting Chang et al.
American journal of physiology. Lung cellular and molecular physiology, 302(11), L1159-L1166 (2012-03-27)
The pathophysiology of congenital diaphragmatic hernia (CDH) is constituted by pulmonary hypoplasia and pulmonary hypertension (PH). We previously reported successful treatment with imatinib of a patient with CDH. This study examines the effect of antenatal imatinib administration on the pulmonary
Yuriko Watanabe et al.
Pediatric research, 73(3), 344-348 (2013-01-25)
We examined the extent of fetal lung hypoplasia and lung maturation using the amniotic lamellar body count (LBC) in congenital diaphragmatic hernia (CDH). We obtained 30 amniotic fluid samples from human CDH cases during cesarean section. We assessed LBC, magnetic
Florian Friedmacher et al.
Pediatric surgery international, 29(1), 19-24 (2012-11-13)
The high morbidity of newborn infants with congenital diaphragmatic hernia (CDH) is attributed to pulmonary hypoplasia (PH), which is characterized by a failure of alveolar development. The nitrofen-induced CDH model has been widely used to investigate the pathogenesis of PH
J J Greer et al.
Pediatric pulmonology, 29(5), 394-399 (2000-05-02)
In this review, we discuss recent advances in the study of the pathogenesis of congenital diaphragmatic hernia (CDH). Much of the research has involved the use of an animal model of CDH in which diaphragmatic defects are produced in fetal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service