Skip to Content
Merck
All Photos(1)

Key Documents

33307

Supelco

Sulfosulfuron

PESTANAL®, analytical standard

Synonym(s):

1-(4,6-Dimethoxypyrimidin-2-yl)-3-(2-ethylsulfonylimidazol[1,2-a]pyridin-3-ylsulfonyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N6O7S2
CAS Number:
Molecular Weight:
470.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCS(=O)(=O)c1nc2ccccn2c1S(=O)(=O)NC(=O)Nc3nc(OC)cc(OC)n3

InChI

1S/C16H18N6O7S2/c1-4-30(24,25)13-14(22-8-6-5-7-10(22)17-13)31(26,27)21-16(23)20-15-18-11(28-2)9-12(19-15)29-3/h5-9H,4H2,1-3H3,(H2,18,19,20,21,23)

InChI key

RBSXHDIPCIWOMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfosulfuron is a pre- and post-emergence sulfonyl urea herbicide, widely used in controlling grass and broad-leaf weeds in crop production. It is found to be highly effective against O.aegyptiaca.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfosulfuron may be used as an analytical reference standard for the determination of the analyte in water samples using solid-phase extraction and ultra-high-pressure liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Trace level determination of selected sulfonylurea herbicides in wetland sediment by liquid chromatography electrospray tandem mass spectrometry.
Deganhardt D, et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 45(1), 11-24 (2009)
Rapid determination of sixteen sulfonylurea herbicides in surface water by solid phase extraction cleanup and ultra-high-pressure liquid chromatography coupled with tandem mass spectrometry.
Yan C, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879(30), 3484-3489 (2011)
Dan-Dan Wang et al.
Journal of chromatographic science, 59(1), 95-102 (2020-10-21)
A novel magnetic solid-phase extraction technique coupled to ultraperformance liquid chromatography has been developed for separation and preconcentration of four sulfonylurea herbicides (sulfosulfuron, bensulfuron-methyl, pyrazosulfuron-ethyl and halosulfuro-methyl) in aqueous samples. The key point of this method was the application of
Amit Paporisch et al.
Pest management science, 77(1), 253-263 (2020-07-21)
Residual herbicides are an important component in many weed control strategies. Their herbicidal activity depends on their fate in soil, with respect to the required concentration for weed control in space and time. In this study, the effect of weather
Mohammad Mehdizadeh et al.
Bulletin of environmental contamination and toxicology, 99(1), 93-99 (2017-04-05)
A HPLC-UV detection system was used for determination of sulfosulfuron and tribenuron methyl residues from soils. The soils were fortified with sulfosulfuron and tribenuron methyl at rates of 26 and 15 g a.i. ha

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service