Skip to Content
Merck
All Photos(1)

Key Documents

32974

Supelco

Forchlorfenuron

PESTANAL®, analytical standard

Synonym(s):

N-(2-Chloro-4-pyridyl)-N′-phenylurea, 4-CPPU, CPPU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10ClN3O
CAS Number:
Molecular Weight:
247.68
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cc(NC(=O)Nc2ccccc2)ccn1

InChI

1S/C12H10ClN3O/c13-11-8-10(6-7-14-11)16-12(17)15-9-4-2-1-3-5-9/h1-8H,(H2,14,15,16,17)

InChI key

GPXLRLUVLMHHIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Forchlorfenuron is a synthetic cytokinin and a growth hormone which accelerates cell division and promotes organ formation and protein synthesis.

Application

Forchlorfenuron may be used as a reference standard for the determination of forchlorfenuron, a plant growth regulator in fruits by modified QuEChERS procedure and UPLC−MS/MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of forchlorfenuron and abscisic acid on the quality of `Flame Seedless? grapes.
Peppi MC and Fidelibus MW
Hortscience: a Publication of the American Society For Horticultural Science Hortscience, 43(1), 173-176 (2008)
Simultaneous determination of plant growth regulators in fruits using a modified QuEChERS procedure and UPLC-MS/MS.
Yan Z, et al.
Horticultural Plant Journal, 2(4), 203-208 (2016)
Forchlorfenuron detection based on its inhibitory effect towards catalase immobilized on boron nitride substrate.
Xu Q, et al.
Biosensors And Bioelectronics, 63(1), 294-300 (2015)
F Q Fu et al.
Plant biology (Stuttgart, Germany), 12(1), 98-107 (2010-07-27)
We investigated the temporal and spatial changes in cell division, endoreduplication and expression of cell cycle-related genes in developing cucumber fruits at 0-20 days after anthesis (DAA). Cell division was intense at 0-4 DAA and then decreased until to 8
Guoying Wang et al.
Journal of plant physiology, 163(6), 591-600 (2005-12-07)
It has previously been shown that boron (B) deficiency inhibits growth of the plant apex, which consequently results in a relatively weak apical dominance, and a subsequent sprouting of lateral buds. Auxin and cytokinins (CKs) are the two most important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service