Skip to Content
Merck
All Photos(2)

Key Documents

32753

Supelco

Cyhalofop-butyl

PESTANAL®, analytical standard, mixture of stereoisomers

Synonym(s):

Butyl (R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20FNO4
CAS Number:
Molecular Weight:
357.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOC(=O)[C@@H](C)Oc1ccc(Oc2ccc(cc2F)C#N)cc1

InChI

1S/C20H20FNO4/c1-3-4-11-24-20(23)14(2)25-16-6-8-17(9-7-16)26-19-10-5-15(13-22)12-18(19)21/h5-10,12,14H,3-4,11H2,1-2H3/t14-/m1/s1

InChI key

TYIYMOAHACZAMQ-CQSZACIVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyhalofop-butyl is a post-emergence, aryloxyphenoxypropionate herbicide, which is used in controlling weeds in rice plantations. Its mode of action involves the inhibition of acetyl-coenzyme A carboxylase (ACCase) biosynthesis.

Application

Cyhalofop-butyl may be used as a reference standard in the determination of cyhalofop-butyl in rice samples using liquid chromatography coupled with diode array detector (LC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Comparison of extraction solvents and conditions for herbicide residues in milled rice with liquid chromatography-diode array detection analysis (LC-DAD)
Niell.S, et al.
Food Additives and Contaminants, 27(2), 206-211 (2010)
Basis of selectivity of cyhalofop-butyl in Oryza sativa L.
Ruiz-Santaella.PJ, et al.
Planta, 223(2), 191-199 (2006)
Barnyardgrass (Echinochloa crus-galli) control in water-seeded rice (Oryza sativa) with cyhalofop-butyl
Ntanos.AD, et al.
Weed Technology, 14(2), 383-388 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service