Skip to Content
Merck
All Photos(1)

Key Documents

32539

Supelco

Nodularin solution

10 μg/mL in methanol, analytical standard

Synonym(s):

Cyclo[(2S,3S,4E,6E,8S,9S)-3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-4,6-decadienoyl-D-γ-glutamyl-(2Z)-2-(methylamino)-2-butenoyl-(3S)-3-methyl-Dβ-aspartyl-L-arginyl), Nodularin-R solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H60N8O10
CAS Number:
Molecular Weight:
824.96
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)[C@@H](NC(=O)\C(=C\C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C41H60N8O10/c1-8-31-38(54)48-34(40(57)58)26(5)36(52)46-29(15-12-20-44-41(42)43)37(53)45-28(25(4)35(51)47-30(39(55)56)18-19-33(50)49(31)6)17-16-23(2)21-24(3)32(59-7)22-27-13-10-9-11-14-27/h8-11,13-14,16-17,21,24-26,28-30,32,34H,12,15,18-20,22H2,1-7H3,(H,45,53)(H,46,52)(H,47,51)(H,48,54)(H,55,56)(H,57,58)(H4,42,43,44)/b17-16+,23-21+,31-8-/t24-,25-,26-,28-,29-,30+,32-,34+/m0/s1

InChI key

IXBQSRWSVIBXNC-HSKGSTCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nodularins belong to the class of hepatotoxic cyclic pentapeptides and have their structures resembling to that of microcystins. They are produced by Nodularia spumigena cyanobacterium.

Application

Nodularin may be used as an analytical reference standard for the determination of the analyte in:
  • Surface and drinking waters by on-line solid phase extraction (SPE) and ultra high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).
  • Sediments and pore waters by accelerated solvent extraction (ASE) and HPLC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Al-Tebrineh et al.
Toxicon : official journal of the International Society on Toxinology, 57(4), 546-554 (2011-01-05)
Toxin-producing cyanobacteria are a worldwide threat to both human and animal health. The hepatotoxins microcystin and nodularin are the most commonly occurring toxins produced by bloom-forming cyanobacteria. They are cyclic peptides that are synthesized nonribosomally by a multienzyme complexes encoded
Analysis of intracellular and extracellular microcystin variants in sediments and pore waters by accelerated solvent extraction and high performance liquid chromatography-tandem mass spectrometry.
Zastepa A, et al.
Analytica Chimica Acta, 872(1-2), 26-34 (2015)
Andrea Gambaro et al.
Rapid communications in mass spectrometry : RCM, 26(12), 1497-1506 (2012-05-18)
Cyanobacteria are a small group of photosynthetic planktonic bacteria, producing a large group of strong hepatotoxins called microcystins (MCs). Many studies have been conducted to evaluate the presence of MCs and nodularin (NOD) in water or in marine organisms, but
Rebecca A Halvorson et al.
Analytical chemistry, 83(24), 9273-9280 (2011-11-01)
Raman spectra of microcystin-LR (MC-LR), MC-RR, MC-LA, MC-LF, MC-LY, MC-LW, MC-YR, and nodularin collected by drop-coating deposition Raman (DCDR) spectroscopy are sufficiently unique for variant identification. Amino acid spectra of L-phenylalanine, L-leucine, L-alanine, D-alanine, L-glutamic acid, L-arginine, L-tryptophan, L-tyrosine, and
S A Wood et al.
Journal of environmental monitoring : JEM, 13(6), 1617-1624 (2011-04-15)
The cyanobacterial toxins, nodularin and microcystin, are highly efficient inhibitors of cellular protein phosphatases. Toxicity primarily evolves following ingestion of cyanobacterial material or toxins and results in liver and renal pathology. Ingestion is the main route of exposure in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service