Skip to Content
Merck
All Photos(2)

Key Documents

32436

Supelco

Ofloxacin-d3

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D3H17FN3O4
CAS Number:
Molecular Weight:
364.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N1CCN(CC1)c2c(F)cc3C(=O)C(=CN4C(C)COc2c34)C(O)=O

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/i2D3

InChI key

GSDSWSVVBLHKDQ-BMSJAHLVSA-N

General description

Ofloxacin-d3 is a deuterated labeled compound typically used as an analytical standard in chromatography applications.

Application

Ofloxacin-d3 may be used as an analytical standard for the determination of ofloxacin in:
  • Hospital wastewater samples by ultra-performance liquid chromatography-electrospray ionization-tandem mass spectrometry (UPLC-ESI-MS/MS) with multiple reaction monitoring (MRM) detection.
  • Raw bovine milk and commercial skimmed milk samples as well as bottled natural mineral waters by solid phase extraction (SPE) and LC-ESI-MS/MS operating in (MRM) mode of detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid residue analysis of fluoroquinolones in raw bovine milk by online solid phase extraction followed by liquid chromatography coupled to tandem mass spectrometry.
Kantiani L, et al.
Journal of Chromatography A, 1218(50), 9019-9027 (2011)
Factors influencing the extraction of pharmaceuticals from sewage sludge and soil: an experimental design approach.
Ferhi S, et al.
Analytical and Bioanalytical Chemistry, 408(22), 6153-6168 (2016)
Analysis of multiclass antibiotic residues in urban wastewater in Tunisia.
Harrabi M, et al.
Environmental Nanotechnology, Monitoring & Management, 10(22), 163-170 (2018)
Ultra-trace analysis of hormones, pharmaceutical substances, alkylphenols and phthalates in two French natural mineral waters.
Devier MH, et al.
The Science of the Total Environment, 443(50), 621-632 (2013)
Calibration and field evaluation of polar organic chemical integrative sampler (POCIS) for monitoring pharmaceuticals in hospital wastewater.
Bailly E, et al.
Environmental Pollution (Barking, Essex : 1987), 174(22), 100-105 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service