Skip to Content
Merck
All Photos(1)

Key Documents

29135

Supelco

Cyclohexanone

Selectophore, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for ion-selective electrodes

Quality Level

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

description

solvent for preparation of membranes

product line

Selectophore

Assay

≥99.5% (GC)
≥99.5%

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

impurities

≤0.1% water

refractive index

n20/D 1.450 (lit.)
n20/D 1.451

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanone is an industrially important reagent. It is also a major intermediate in the synthesis of nylon.[1]
Visit our Sensor Applications portal to learn more.

Other Notes

Solvent for preparing ion-selective membranes for microelectrodes; it is less volatile than other solvents[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U. Schaller et al.
Pflugers Archiv : European Journal of Physiology (1993)
Selective phenol hydrogenation to cyclohexanone over a dual supported Pd-Lewis acid catalyst.
Liu H
Science, 326 (5957), 1250-1252 (2009)
Effect of hydrogen sulphide on the hydrodeoxygenation of aromatic and aliphatic oxygenates on sulphided catalysts.
Senol OI
J. Mol. Catal. A: Chem., 277 (1), 107-112 (2007)
Won-Heong Lee et al.
Applied microbiology and biotechnology, 97(4), 1561-1569 (2012-10-12)
Sufficient supply of NADPH is one of the most important factors affecting the productivity of biotransformation processes. In this study, construction of an efficient NADPH-regenerating system was attempted using direct phosphorylation of NADH by NADH kinase (Pos5p) from Saccharomyces cerevisiae
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2638-2643 (2012-09-07)
The processing of renewable feedstocks to platform chemicals and, to a lesser degree, fuels is a key part of sustainable development. In particular, the combination of lignocellulosic biomass with hydrothermal upgrading (HTU), using high temperature and pressure water (HTPW), is

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service