Skip to Content
Merck
All Photos(1)

Key Documents

25508

Sigma-Aldrich

Potassium sodium tartrate tetrahydrate

puriss., 99.0-101.0% (calc. on H2O free substance), meets analytical specification of Ph. Eur., BP, USP, FC, E 337

Synonym(s):

L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOCOCH(OH)CH(OH)COONa · 4H2O
CAS Number:
Molecular Weight:
282.22
Beilstein:
6113568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.0-101.0% (calc. on H2O free substance)
form:
solid
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

99.0-101.0% (calc. on H2O free substance)

form

solid

optical activity

[α]20/D +28.0 to +30.0°

quality

meets analytical specification of Ph. Eur., BP, USP, FC, E 337

impurities

acidity or alkalinity, in accordance
residual solvent, complies
≤0.001% heavy metals (as Pb)
≤0.003% total nitrogen (N)
≤0.004% ammonium (NH4)
≤0.004% free alkali (as NaOH)
≤0.0075% free acid (as C4H6O6)
24-26% water (Karl Fischer)

loss

21-26% loss on drying, 150 °C, 3 h

pH

6.5-8.5 (20 °C, 1%)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤1 mg/kg
Ba:, in accordance
Ca: ≤50 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Zn: ≤10 mg/kg

suitability

passes test for appearance of solution

SMILES string

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1

InChI key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium sodium tartrate tetrahydrate (Rochelle salt) exists in the ferroelectric phase in the temperature range of 255 to 297K. In this phase, it crystallizes in the monoclinic system and exhibits space group P21. Saturated aqueous solution of potassium sodium tartrate (Rochelle salt) has been used for the preparation of its tetrahydrate crystals.[1]
Potassium sodium tartrate tetrahydrate is a ferroelectric crystal[2] with a high piezoelectric effect and electromechanical coupling coefficient.[3]

Application

Potassium sodium tartrate tetrahydrate has been used in the β-1,3-glucanase activity assay.[4]
Potassium sodium tartrate tetrahydrate may be used as a component to prepare DNS (3,5- dinitrosalicylic acid) reagent and Fehling′s solution B, which are used in the determination of reducing sugar.[5][6]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan T, et al
Fundamentals of Piezoelectric Sensorics: Mechanical, Dielectric, and Thermodynamical Properties of Piezoelectric Materials, 11-11 (2010)
Wen-Bin Du et al.
Analytical chemistry, 82(23), 9941-9947 (2010-11-04)
This paper describes DropLab, an automated microfluidic platform for programming droplet-based reactions and screening in the nanoliter range. DropLab can meter liquids with picoliter-scale precision, mix multiple components sequentially to assemble composite droplets, and perform screening reactions and assays in
Screening of African traditional vegetables for their alpha-amylase inhibitory effect.
Odhav B, et al.
Journal of Medicinal Plants Research, 4(14), 1502-1507 (2010)
Potassium sodium (2R, 3R)-tartrate tetrahydrate: the paraelectric phase of Rochelle salt at 105K.
Gorbitz CH and Sagstuen E.
Acta Crystallographica Section E, Structure Reports Online, 64(4), 507-508 (2008)
Nandini P Shetty et al.
Journal of experimental botany, 60(15), 4287-4300 (2009-11-03)
The accumulation of the pathogenesis-related (PR) proteins beta-1,3-glucanase and chitinase and structural defence responses were studied in leaves of wheat either resistant or susceptible to the hemibiotrophic pathogen Septoria tritici. Resistance was associated with an early accumulation of beta-1,3-glucanase and

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service