Skip to Content
Merck
All Photos(3)

Key Documents

247367

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H,23H-porphine

≥99%

Synonym(s):

meso-Tetraphenylporphyrin, TPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H30N4
CAS Number:
Molecular Weight:
614.74
Beilstein:
379542
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥99%

form

crystalline powder

impurities

<0.1% corresponding chlorin

mp

>300 °C (lit.)

solubility

toluene: 0.03 g/L, clear

λmax

415 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

c1ccc(cc1)-c2c3ccc(n3)c(-c4ccccc4)c5ccc([nH]5)c(-c6ccccc6)c7ccc(n7)c(-c8ccccc8)c9ccc2[nH]9

InChI

1S/C44H30N4/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36/h1-28,45,48H/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-

InChI key

YNHJECZULSZAQK-LWQDQPMZSA-N

Gene Information

human ... TERT(7015)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zerrin Sezgin et al.
International journal of pharmaceutics, 332(1-2), 161-167 (2006-10-24)
Meso-tetraphenyl porphine (mTPP) is a highly lipophilic, fluorescent porphyrin derivate and it is used as photosensitizer on the treatment of malign neoplasms. The aim of this study was to prepare mTPP loaded pluronic F127 and polyethylene glycol-distearoyl phosphatidylethanolamine (PEG(2000)-DSPE) micelles
Anette Weyergang et al.
Biochimica et biophysica acta, 1830(3), 2659-2670 (2013-05-15)
Tyrosin kinase inhibitors (TKIs) and monoclonal antibodies aimed to target epidermal growth factor receptor (EGFR) have shown limited effect as monotherapies and drug resistance is a major limitation for therapeutic success. Adjuvant therapies to EGFR targeting therapeutics are therefore of
Maria Bassiouk et al.
Journal of nanoscience and nanotechnology, 11(6), 5457-5468 (2011-07-21)
The self-assembly of porphyrins into highly organized functional arrays supported on appropriate solid substrates is an area of research with multiple potential applications in the "bottom-up" approach to manufacturing. In order to analyze the self-assembly of meso-tetraphenylporphine (H2TPP) on the
Satoshi Tsukahara et al.
Analytical and bioanalytical chemistry, 395(4), 1047-1053 (2009-07-09)
Rhombic-ordered microdomains of diprotonated 5,10,15,20-tetraphenylporphine aggregate, whose sizes were 10-200 microm, were formed at dodecane/aqueous H(2)SO(4) interfaces. The light excitation of their two absorption bands (410 and 473 nm for H- and J-bands, respectively) led to one fluorescence band at
Zachary J Tonzetich et al.
Inorganic chemistry, 50(4), 1570-1579 (2011-01-20)
Several nitrosyl complexes of Fe and Co have been prepared using the sterically hindered Ar-nacnac ligand (Ar-nacnac = anion of [(2,6-diisopropylphenyl)NC(Me)](2)CH). The dinitrosyliron complexes [Fe(NO)(2)(Ar-nacnac)] (1) and (Bu(4)N)[Fe(NO)(2)(Ar-nacnac)] (2) react with [Fe(III)(TPP)Cl] (TPP = tetraphenylporphine dianion) to generate [Fe(II)(NO)(TPP)] and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service