Skip to Content
Merck
All Photos(4)

Key Documents

230553

Sigma-Aldrich

Sodium ethoxide solution

21 wt. % in ethanol

Synonym(s):

Sodium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2ONa
CAS Number:
Molecular Weight:
68.05
Beilstein:
3593646
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

form

liquid

concentration

21 wt. % in ethanol

refractive index

n20/D 1.385

density

0.868 g/mL at 25 °C

SMILES string

[Na+].CC[O-]

InChI

1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

QDRKDTQENPPHOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium ethoxide is an alkoxide salt mainly used as a strong base in organic reactions such as deprotonation, dehydration and dehalogenation.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1A - STOT SE 2

Target Organs

Eyes,Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Ethoxide
Whitaker KS and Whitaker DT
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
S H Brorson
Micron (Oxford, England : 1993), 32(2), 101-105 (2000-08-11)
The study's purpose was to obtain improved "deplasticizing" of epoxy sections for immunoelectron microscopy. Epoxy-embedded renal swine tissue with immune complex deposits was used. Ultrathin sections were mounted on uncoated grids or on carbon-stabilized formvar grids. The sections were exposed
Francesca Monticelli et al.
Journal of endodontics, 32(5), 447-451 (2006-04-25)
The influence of different silane/dentin adhesives on the microtensile bond strengths of a hybrid composite to sodium ethoxide-etched quartz fiber posts was investigated. A prehydrolyzed silane was applied without an adhesive and compared to two-component systems in which hydrolysis of
S H Brorson
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 45(1), 143-146 (1997-01-01)
The purpose of this investigation was to develop a method that could be used to estimate how damaging sodium ethoxide is to different antigens with respect to immunolabeling when epoxy sections are deplasticized. If we obtain weak labeling for an
Ali Reza Harifi-Mood et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 7(1), 88-91 (2004-05-18)
Variable-Temperature Kinetics has been used to obtain the rate constants of the reaction at various temperatures during one kinetic run. Pseudo-first-order rate constants for the transesterification of procaine with aliphatic alcohols ethanol, n-propanol and tert-butanol were obtained by the fluorescence

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service