Skip to Content
Merck
All Photos(1)

Key Documents

17774

Millipore

Potassium tellurite solution

1% in H2O, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

Agency

according to ISO 6888-1:2020

Quality Level

sterility

sterile (Filtered and Aseptic Handled)

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

1% in H2O

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Corynebacterium spp.
Staphylococcus spp.

SMILES string

[K+].[K+].[O-][Te]([O-])=O

InChI

1S/2K.H2O3Te/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BFPJYWDBBLZXOM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

A filter sterilized solution of potassium tellurite for the selective isolation of Staphylococci and Corynebacteria.

Other Notes

10 mls per vial
sterile 1% Potassium tellurite Solution in water

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel E Castro et al.
Biochemical and biophysical research communications, 380(1), 148-152 (2009-01-27)
Potassium tellurite (K(2)TeO(3)) is harmful to most organisms and specific mechanisms explaining its toxicity are not well known to date. We previously reported that the lpdA gene product of the tellurite-resistant environmental isolate Aeromonas caviae ST is involved in the
Miguel E Castro et al.
Biochemical and biophysical research communications, 375(1), 91-94 (2008-08-05)
Potassium tellurite (K(2)TeO(3)) is extremely toxic for most forms of life and only a limited number of organisms are naturally resistant to the toxic effects of this compound. Crude extracts prepared from the environmental isolate Aeromonas caviae ST catalize the
Lillian G Acuña et al.
Archives of microbiology, 191(5), 473-476 (2009-03-31)
Potassium tellurite is highly toxic to most forms of life and specific bacterial tellurite defense mechanisms are not fully understood to date. Recent evidence suggests that tellurite would exert its toxic effects, at least in part, through the generation of
C C Vásquez et al.
Current microbiology, 43(6), 418-423 (2001-10-31)
The nucleotide sequence of a 4,539 bp fragment of Bacillus stearothermophilus V mediating tellurite resistance in Escherichia coli was determined. Four ORFs of more than 150 amino acids encoding polypeptides of 244, 258, 308, and 421 residues were found in
Domenica R Massardo et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 22(6), 1089-1094 (2009-09-18)
The effects of potassium tellurite on growth and survival of rho(+) and rho(0) Saccharomyces cerevisiae strains were investigated. Both rho(+) and rho(0) strains grew on a fermentable carbon source with up to 1.2 mM K(2)TeO(3), while rho(+) yeast cells grown

Articles

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service