Skip to Content
Merck
All Photos(1)

Key Documents

16377

Millipore

MacConkey Broth Purple

suitable for microbiology, NutriSelect® Plus

Synonym(s):

MacConkey Bromocresol purple Broth, MacConkey G Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bile salts, 5.0 g/L
bromoresol purple, 0.01 g/L
lactose, 10.0 g/L
peptone, 20.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Citrobacter spp.
selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Serratia spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Looking for similar products? Visit Product Comparison Guide

Application

MacConkey Broth Purple is a differential medium used for the detection of coliforms in water samples and milk examination by MPN technique. It is a modification of the classic medium, where neutral red is replaced by a less aggressive indicator, according European Pharmacopoeia. It is also recommended by ISO Committee under the specification ISO 9308-2:1990.

Preparation Note

Suspend 40 g in 1000 ml distilled water. Dispense in test tubes with inverted Durham tubes. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service