Skip to Content
Merck
All Photos(1)

Key Documents

150495

Sigma-Aldrich

Isopentyl nitrite

96%

Synonym(s):

Isoamyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2ONO
CAS Number:
Molecular Weight:
117.15
Beilstein:
969510
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

96%

form

liquid

technique(s)

titration: suitable

color

colorless to green-yellow
colorless to yellow

refractive index

n20/D 1.386 (lit.)

bp

99 °C (lit.)

mp

<-80 °C

solubility

water: soluble 0.38 g/L at 25 °C

density

0.872 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CC(C)CCON=O

InChI

1S/C5H11NO2/c1-5(2)3-4-8-6-7/h5H,3-4H2,1-2H3

InChI key

OWFXIOWLTKNBAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopentyl nitrite, also known as isoamyl nitrite, is an oxidant, and nitrosating agent for phosphoramidites which rearrange into phosphotriesters during nitrosation.

Application

Isopentyl nitrite has been used:
  • as a radical initiator to functionalize multi-walled carbon nanotubes.
  • as a reagent in the preparation of 4-carboxy-phenyl diazonium salt tetrafluoroborate.
  • in the synthesis of peroxynitrite (ONOO-).
Isopentyl nitrite, also known as isoamyl nitrite, is used for the generation of benzyne from anthranilic acid. It is a smooth muscle relaxant that may be suitable as nitrovasodilators and as recreational drugs known as poppers. Its applications are also seen in kits to treat cyanide poisoning, its function being to oxidize hemoglobin (Hb) to methemoglobin (metHb) which reacts with cyanide to form the non-toxic cyano-metHb.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron transfer at Au surfaces modified by Tethered Osmium bipyridine?pyridine complexes
Journal of Solid State Electrochemistry, 11:11, 1511-1520 null
New insights into the functionalization of multi-walled carbon
nanotubes with aniline derivatives
Lipinska M E
Carbon, 50, 3280 -33294 (2012)
Comparing nitrosative versus oxidative stress toward zinc finger-dependent transcription. Unique role for NO.
Kroncke KD
The Journal of Biological Chemistry, 277(15), 13294-13301 (2002)
Tetrahedron Letters, 31, 1113-1113 (1990)
R M Uppu et al.
Analytical biochemistry, 236(2), 242-249 (1996-05-01)
A new method for the preparation of high concentrations of peroxynitrite (up to 1 M) is described. The synthesis uses a two-phase system and involves a displacement reaction by the hydroperoxide anion (in the aqueous phase) on isoamyl nitrite (in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service