Skip to Content
Merck
All Photos(2)

Key Documents

14659

Supelco

Potassium phthalate monobasic

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

Standard for quantitative NMR
certified reference material
TraceCERT®

Quality Level

description

qNMR Standard for D2O (8.2 ppm / 7.5 ppm)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable
qNMR: suitable

pH

4

mp

295-300 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)

Other Notes

Chemical Shift: ~7-8 ppm (chemical shifts may slightly vary depending on the experimental conditions)
suitable NMR solvents: D2O, MeOD

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Hanne Frederiksen et al.
Environmental science & technology, 47(2), 958-967 (2012-12-14)
Urinary phthalate excretion is used as marker of phthalate exposure in epidemiological studies. Here we examine the reliability of urinary phthalate levels in exposure classification by comparing the inter- and intrasubject variation of urinary phthalate metabolite levels. Thirty-three young healthy
Juanjuan Feng et al.
Journal of chromatography. A, 1268, 16-21 (2012-11-07)
Because of the occurrence of ion exchange between high-ionic-strength solution and anions of polymeric ionic liquids (PILs), PILs based solid-phase microextraction (SPME) fibers were rarely used in direct immersion mode to high-salt-added samples. In this work, a novel double-confined PIL
Wei He et al.
The Science of the total environment, 461-462, 672-680 (2013-06-19)
The spatio-temporal distributions and the ecological and health risks of PAEs in surface water of Lake Chaohu, the fifth largest lake in China, were studied based on the monthly monitoring of six PAE congeners from May 2010 to April 2011.
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional

Articles

The signal ratio of two different protons can be measured with tremendous precision, which enables the generation of certified reference materials for use as qNMR standards.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

NMR shift charts assist in identifying impurities in deuterated solvents, aiding accurate chemical analysis.

See All

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service