Skip to Content
Merck
All Photos(2)

Key Documents

14432

Millipore

Tryptic Soy Agar - Dehydrated Culture Media

NutriSelect® Plus, (Vegitone), powder, suitable for microbiology

Synonym(s):

CASO Agar, Vegitone, Soybean Casein digest Agar, Vegitone, TSA, Vegitone, Tryptone Soya Agar, Vegitone, Vegitone Tryptic Soy Agar, CASO Agar, Soybean Casein digest Agar, TSA, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

biological source

(Vegitone)

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
papaic digest of soyabean meal, 5.0 g/L
sodium chloride, 5.0 g/L
tryptone (vegetable), 15.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
industrial hygiene
personal care
pharmaceutical
surface monitoring
veterinary

microbiology

suitability

Enterococcus spp.
Haemophilus spp.
Pneumococcus spp.
nonselective for Bacillus spp.
nonselective for Bacteroides spp.
nonselective for Campylobacter spp.
nonselective for Candida spp.
nonselective for Escherichia coli
nonselective for Neisseria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

Application

Vegitone Tryptic Soy Agar is a general purpose medium used for cultivation of a wide variety of microorganisms. It is free of any material derived from animals.

Preparation Note

Suspend 40.0 g in 1000 ml distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. If desired, aseptically add 5% v/v defibrinated blood in previously cooled medium (45-50°C).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service