Skip to Content
Merck
All Photos(4)

Key Documents

142387

Sigma-Aldrich

2,4,6-Trimethylpyridine

ReagentPlus®, 99%

Synonym(s):

2,4,6-Collidine, sym-Collidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Beilstein:
107283
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

171-172 °C (lit.)

mp

−43 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)nc(C)c1

InChI

1S/C8H11N/c1-6-4-7(2)9-8(3)5-6/h4-5H,1-3H3

InChI key

BWZVCCNYKMEVEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Trimethylpyridine is a pyridine derivative. It has a pK of 7.4. The product can react with trifluoroiodomethane in cyclopentane solution to afford 1:1 complex. This complex was investigated by NMR (Nuclear Magnetic Resonance) spectroscopy. Collidine-buffered osmium tetroxide solutions have been prepared by adding osmium tetroxide solution to it. These solutions have been used as fixative for electron microscopic studies.
2,4,6-Trimethylpyridine can undergo oxidation with potassium permanganate to form 2,4,6-pyridinetricarboxylic acid.
Hindered base.

Application

2,4,6-Trimethylpyridine (collidine, s-collidine, 2,4,6-collidine) has been used in the preparation of a library of N-acyl aminocoumarins bearing low-molecular-weight N-acyl groups.
Tissue fixative for electron microscopy.
Useful solvent for the cleavage of hindered esters by anhydrous lithium iodide.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, photoluminescence and magnetic properties of new 3-D lanthanide-pyridine-2, 4, 6-tricarboxylate frameworks
Li C-J, et al.
CrystEngComm, 10(11), 1645-1652 (2008)
Janet E Del Bene et al.
The journal of physical chemistry. A, 110(3), 1128-1133 (2006-01-20)
Ab initio EOM-CCSD calculations have been performed on 3:1 FH:NH3 complexes at their own optimized MP2/6-31+G(d,p) geometries and at the optimized geometries in the hydrogen-bonding regions of corresponding 3:1 FH:collidine complexes. The isolated gas-phase equilibrium 3:1 FH:NH3 complex has an
Leszek Pazderski et al.
Magnetic resonance in chemistry : MRC, 48(6), 417-426 (2010-05-18)
(1)H, (13)C and (15)N NMR studies of gold(III), palladium(II) and platinum(II) chloride complexes with dimethylpyridines (lutidines: 2,3-lutidine, 2,3lut; 2,4-lutidine, 2,4lut; 3,5-lutidine, 3,5lut; 2,6-lutidine, 2,6lut) and 2,4,6-trimethylpyridine (2,4,6-collidine, 2,4,6col) having general formulae [AuLCl(3)], trans-[PdL(2)Cl(2)] and trans-/cis-[PtL(2)Cl(2)] were performed and the respective
J L Tomsig et al.
Histochemistry, 89(3), 301-306 (1988-01-01)
In previous work of our laboratory it was demonstrated that collidine (2,4,6-trimethylpyridine) abolishes the core osmiophilia and chromaffin reaction from rat pinal gland and vas deferens nerves. This abolition was apparent when tissues were briefly incubated in collidine or when
Hiromichi Fujioka et al.
Chemistry, an Asian journal, 7(2), 367-373 (2011-12-14)
Mild substitution reactions of acetals with carbon nucleophiles via the pyridinium-type salts generated by the treatment of acetals with TESOTf-2,4,6-collidine or 2,2'-bipyridyl have been developed. Various carbon nucleophiles, such as organocuprates, silyl enol ethers, enamines, etc., reacted with the pyridinium-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service