Skip to Content
Merck
All Photos(1)

Key Documents

11569

Supelco

3,4-Dihydroxyphenylacetic acid

analytical standard

Synonym(s):

DOPAC, Homoprotocatechuic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein:
2211017
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)
97.0-103.0% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

127-130 °C (lit.)

application(s)

pharmaceutical

format

neat

SMILES string

OC(=O)Cc1ccc(O)c(O)c1

InChI

1S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)

InChI key

CFFZDZCDUFSOFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dihydroxyphenylacetic acid may be used as a reference standard for the determination of 3,4-dihydroxyphenylacetic acid in rat samples by high-performance liquid chromatography with electrochemical detection.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 3, 4?dihydroxyphenylalanine, 5?hydroxytryptophan, dopamine, 4?hydroxy?3?methoxyphenylalanine, norepinephrine, 3, 4?dihydroxyphenylacetic acid, homovanillic acid, serotonin, and 5?hydroxyindoleacetic acid in rat cerebrospinal fluid and brain by high?performance liquid chromatography with electrochemical detection.
Wagner J, et al.
Journal of Neurochemistry, 38(5), 1241-1254 (1982)
Xiaoli Liu et al.
Neurotoxicity research, 38(1), 27-37 (2020-03-22)
Parkinson's disease (PD) is a neurodegenerative disorder characterized by the accumulation of Lewy bodies and loss of dopaminergic neurons in the substantia nigra pars compacta (SNpC). MPTP is widely used to generate murine PD model. In addition to classical motor
Jinhee Kim et al.
Biomolecules & therapeutics, 28(3), 282-291 (2020-04-28)
Inhaled solvents such as toluene are of particular concern due to their abuse potential that is easily exposed to the environment. The inhalation of toluene causes various behavioral problems, but, the effect of short-term exposure of toluene on changes in
Kangwei Li et al.
Journal of ethnopharmacology, 250, 112487-112487 (2019-12-21)
Seahorses (Hippocampus erectus), belonging to syngnathidae of syngnathiformes, are a traditional Chinese medicine for increasing and balancing vital energy within the body and brain, as well as calming mood and improving sleep. Based on the hypothesis of monoamine neurotransmitter deficiency
Wanqiu Huang et al.
Journal of neurochemistry, 150(3), 282-295 (2019-05-24)
Neuroinflammation is one of the driving forces of progressive neurodegeneration in Parkinson's disease (PD). The metabolomics approach has been proved highly useful in identifying potential therapeutic targets. Here, to identify inflammation-relevant treatment targets for PD, mass spectrometry-based untargeted metabolomics was

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service