Skip to Content
Merck
All Photos(2)

Documents

104043

Sigma-Aldrich

N-Phenyl-1-naphthylamine

reagent grade, 98%

Synonym(s):

1-(N-phenylamino)naphthalene, N-(1-Naphthyl)aniline, NPN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHC6H5
CAS Number:
Molecular Weight:
219.28
Beilstein:
2211174
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

Assay

98%

form

solid

bp

226 °C/15 mmHg (lit.)

mp

60-62 °C (lit.)

λmax

252 nm

SMILES string

N(c1ccccc1)c2cccc3ccccc23

InChI

1S/C16H13N/c1-2-9-14(10-3-1)17-16-12-6-8-13-7-4-5-11-15(13)16/h1-12,17H

InChI key

XQVWYOYUZDUNRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Phenyl-1-naphthylamine can be used as fluorescent probe for the determination of critical micelle concentration of surfactants. N-Phenyl-1-naphthylamine was used in a method for determination of the concentration of organolithium and organomagnesium reagents. N-Phenyl-1-naphthylamine was used as hydrophobic probe to study the phase transitions of membrane lipids in whole cells .

Biochem/physiol Actions

N-Phenyl-1-naphthylamine turns fluorescent after binding to hydrophobic regions of cell membranes.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sasitorn Chusri et al.
The Journal of antimicrobial chemotherapy, 64(6), 1203-1211 (2009-10-29)
The emergence of antibiotic resistance has seriously diminished antibiotic efficacy and an increasing number of infections are becoming difficult to treat. One approach to the restoration of antibiotic activity is to administer them in conjunction with non-antibiotic compounds that depress
Hanna-Leena Alakomi et al.
Journal of agricultural and food chemistry, 55(10), 3905-3912 (2007-04-19)
Gram-negative bacteria are important food spoilage and pathogenic bacteria. Their unique outer membrane (OM) provides them with a hydrophilic surface structure, which makes them inherently resistant to many antimicrobial agents, thus hindering their control. However, with permeabilizers, compounds that disintegrate
Markus A Seeger et al.
Nature structural & molecular biology, 15(2), 199-205 (2008-01-29)
The AcrA-AcrB-TolC complex is the major multidrug efflux pump in Escherichia coli. The asymmetric structure of the trimeric inner-membrane component AcrB implies functional rotation of the monomers and a peristaltic mode of drug efflux. This mechanism suggests the occurrence of
Guillaume Sautrey et al.
Antimicrobial agents and chemotherapy, 58(8), 4420-4430 (2014-05-29)
The development of novel antimicrobial agents is urgently required to curb the widespread emergence of multidrug-resistant bacteria like colistin-resistant Pseudomonas aeruginosa. We previously synthesized a series of amphiphilic neamine derivatives active against bacterial membranes, among which 3',6-di-O-[(2"-naphthyl)propyl]neamine (3',6-di2NP), 3',6-di-O-[(2"-naphthyl)butyl]neamine (3',6-di2NB)
Xiaoyan Ning et al.
International journal of molecular sciences, 19(2) (2018-02-01)
Glucose oxidase (GOD, EC.1.1.3.4) specifically catalyzes the reaction of β-d-glucose to gluconic acid and hydrogen peroxide in the presence of oxygen, which has become widely used in the food industry, gluconic acid production and the feed industry. However, the poor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service