Skip to Content
Merck
All Photos(1)

Key Documents

06958

Supelco

Butyl acrylate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOO(CH2)3CH3
CAS Number:
Molecular Weight:
128.17
Beilstein:
1749970
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

3.3 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

559 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.0015% hydroquinone monomethyl ether as stabilizer

expl. lim.

9.9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418 (lit.)
n20/D 1.419

bp

145 °C (lit.)

density

0.899 g/mL at 20 °C
0.894 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCOC(=O)C=C

InChI

1S/C7H12O2/c1-3-5-6-9-7(8)4-2/h4H,2-3,5-6H2,1H3

InChI key

CQEYYJKEWSMYFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Junkers et al.
Macromolecular rapid communications, 33(11), 984-990 (2012-05-05)
A novel dithioester control agent [dimethyltetrathioterephtalate (DMTTT)] is presented for the thioketone-mediated radical polymerization (TKMP) of n-butyl acrylate. The rate of polymerization is significantly decreased in the presence of DMTTT indicating formation of dormant radical species. During polymerization, molar masses
Sébastien-Jun Mougnier et al.
Macromolecular rapid communications, 33(8), 703-709 (2012-02-23)
The post-functionalization of poly(3-hexylthiophene) (P3HT) via various synthetic routes is reported. Well-defined and monofunctionalized ω-thiol-terminated P3HT, ω-carboxylic acid-terminated P3HT, ω-acrylate-terminated P3HT, and ω-methacrylate-terminated P3HT are obtained in high yields through a straightforward procedure. From those, different novel P3HT-based graft copolymers
A radio-fluorogenic organic gel for real-time, 3D radiation dosimetry.
John M Warman et al.
Advanced materials (Deerfield Beach, Fla.), 23(42), 4953-4955 (2011-10-13)
Sabrina Hocine et al.
Langmuir : the ACS journal of surfaces and colloids, 29(5), 1356-1369 (2013-01-09)
Thermoresponsive behavior of different kinds of polymersomes was studied using small angle neutron scattering (SANS), transmission electron microscopy (TEM), and proton nuclear magnetic resonance ((1)H NMR). The polymersomes were made of block copolymers containing a 2000 Da polyethylene glycol (PEG)
Yuri Reyes et al.
Macromolecular rapid communications, 32(1), 63-67 (2011-03-25)
Acrylic monomers undergo chain transfer to polymer during polymerization leading to branched and even gelled polymers. It has been experimentally demonstrated that the extent of branching is higher for conventional free radical polymerization than for controlled radical polymerization (ATRP, RAFT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service