Skip to Content
Merck
All Photos(1)

Documents

03951

Sigma-Aldrich

N-Ethylformamide

≥99.0% (GC)

Synonym(s):

N-Formylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHC2H5
CAS Number:
Molecular Weight:
73.09
Beilstein:
1737860
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

100
500

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.432

bp

202-204 °C

density

0.950 g/mL at 20 °C (lit.)

SMILES string

CCNC=O

InChI

1S/C3H7NO/c1-2-4-3-5/h3H,2H2,1H3,(H,4,5)

InChI key

KERBAAIBDHEFDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gil-Soo Han et al.
The Journal of biological chemistry, 283(29), 20443-20453 (2008-05-07)
The Saccharomyces cerevisiae DGK1 gene encodes a diacylglycerol kinase enzyme that catalyzes the formation of phosphatidate from diacylglycerol. Unlike the diacylglycerol kinases from bacteria, plants, and animals, the yeast enzyme utilizes CTP, instead of ATP, as the phosphate donor in
H Cross et al.
Chemical research in toxicology, 3(4), 357-362 (1990-07-01)
Hepatotoxic formamides such as N-methylformamide (NMF) and N,N-dimethylformamide (DMF) are metabolized in vivo to N-acetyl-S-(N-methylcarbamoyl)cysteine via oxidation at the formyl carbon, which yields a reactive intermediate. The hypothesis was tested that this biotransformation route can be studied in vitro with
S P Langdon et al.
Toxicology, 43(3), 239-249 (1987-03-01)
The induction of terminal differentiation in tumour cells represents a possible therapeutic strategy for treating cancer. The alkylformamides are 1 group of experimental compounds which have been shown to induce terminal differentiation in human HL-60 leukemia and murine Friend erythroleukemia
P Kestell et al.
The Journal of pharmacology and experimental therapeutics, 240(1), 265-270 (1987-01-01)
The hepatotoxicity and metabolism of the following close analogs of the hepatotoxic antitumor agent N-methylformamide (NMF) were investigated in CBA/CA mice: N-ethylformamide (NEF), dimethylformamide (DMF), formamide and N-methylacetamide (NMA). Apart from NMF only NEF was potently hepatotoxic as measured by
M R Del Carratore et al.
Environmental and molecular mutagenesis, 36(2), 97-104 (2000-10-03)
A cDNA coding for rat cytochrome P450 2E1 was cloned into the multicopy vector pYeDP60 and expressed in haploid RSY6 and diploid RS112 yeast strains of Saccharomyces cerevisiae under control of the GAL10-CYC1 promoter. Spectral and catalytic properties of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service