Skip to Content
Merck
All Photos(1)

Key Documents

00420580

Harpagoside

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O11
CAS Number:
Molecular Weight:
494.49
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

C[C@@]1(C[C@@H](O)[C@]2(O)C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]12)OC(=O)\C=C\c4ccccc4

InChI

1S/C24H30O11/c1-23(35-16(27)8-7-13-5-3-2-4-6-13)11-15(26)24(31)9-10-32-22(20(23)24)34-21-19(30)18(29)17(28)14(12-25)33-21/h2-10,14-15,17-22,25-26,28-31H,11-12H2,1H3/b8-7+/t14-,15-,17-,18+,19-,20-,21+,22+,23+,24-/m1/s1

InChI key

KVRQGMOSZKPBNS-FMHLWDFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Ju Jeong et al.
European journal of pharmacology, 588(1), 78-84 (2008-05-09)
The cognitive-enhancing activities of E-harpagoside and 8-O-E-p-methoxycinnamoylharpagide (MCA-Hg) isolated from Scrophularia buergeriana were evaluated in scopolamine-induced amnesic mice by the Morris water maze and by passive avoidance tests. E-harpagoside and MCA-Hg significantly improved the impairment of reference memory induced by
Maria Cecilia Anauate et al.
Phytotherapy research : PTR, 24(9), 1365-1369 (2010-09-03)
The present study evaluates the effect of isolated fractions of Harpagophytum procumbens (devil's claw) on cyclooxygenase (COX-1 and COX-2) activities and NO production using a whole blood assay. The activity of COX-1 was quantified as platelet thromboxane B(2) production in
Shengqiang Tong et al.
Phytochemical analysis : PCA, 17(6), 406-408 (2006-12-06)
The bioactive component harpagoside was successfully separated from the crude extract of Scrophularia ningpoensis Hemsley by one-step purification using high-speed counter-current chromatography (HSCCC). A two-phase solvent system containing n-butanol:ethyl acetate:water (1:9:10) was selected following consideration of the partition coefficient of
Cyril Colas et al.
Journal of chromatographic science, 46(2), 174-183 (2008-03-28)
Solid-phase extraction cartridges among those usually used for screening in horse doping analyses are tested to optimize the extraction of harpagoside (HS), harpagide (HG), and 8-para-coumaroyl harpagide (8PCHG) from plasma and urine. Extracts are analyzed by liquid chromatography coupled with
Xiaoyu Sun et al.
Journal of neurochemistry, 120(6), 1072-1083 (2011-12-24)
Parkinson's disease is a chronic neurodegenerative movement disorder characterized by the loss of dopaminergic neurons in the substantia nigra pars compacta. New therapeutic approaches aiming at delaying or reversing the neurodegenerative process are under active investigations. In this work, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service