Skip to Content
Merck
All Photos(1)

Key Documents

00307

Sigma-Aldrich

ADA

BioUltra, ≥99.0% (T)

Synonym(s):

N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
190.15
Beilstein:
1787181
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.1%

loss

≤0.5% loss on drying, 20 °C (HV)

pH

1.5-3.0 (25 °C, 0.01 M in H2O)

useful pH range

6.0-7.2

pKa (25 °C)

6.6

mp

219 °C (dec.) (lit.)

solubility

H2O: 0.05 M, clear, colorless

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.05 M in H2O

UV absorption

λ: 260 nm Amax: 0.20
λ: 280 nm Amax: 0.05

SMILES string

NC(=O)CN(CC(O)=O)CC(O)=O

InChI

1S/C6H10N2O5/c7-4(9)1-8(2-5(10)11)3-6(12)13/h1-3H2,(H2,7,9)(H,10,11)(H,12,13)

InChI key

QZTKDVCDBIDYMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ADA, or N-(2-Acetamido)iminodiacetic acid, can be used to study biological buffers and zwitterionic compounds. ADA has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilised onto the surface of a monolithic capillary column.
Biological buffer component with sodium hydroxide and sodium chloride (pH 5.67-7.57, useful pH range 6.4-7.4). Used to prepare immobilized pH gradients.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birte M Gerken et al.
Analytical chemistry, 77(19), 6113-6117 (2005-10-01)
A novelly developed tweezing-adsorptive bubble separation (ABS) method for the enrichment of metalloenzymes (laccase C and horseradish peroxidase) is introduced. The method is based on the chelation of the enzymes' active center and can also be applied for analysis. N-(2-acetamido)iminodiacetic
Identification of metallothionein isoforms with capillary zone electrophoresis using a polyacrylamide-coated tube
Minami, T., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 685, 353-359 (1996)
Eoin Gillespie et al.
Journal of separation science, 32(15-16), 2659-2667 (2009-07-09)
The application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilised onto the surface of a monolithic capillary column is described. The use of SC(4)D to visualise changes in conductivity
Yin-Cheng Hsieh et al.
The Journal of biological chemistry, 288(42), 30645-30658 (2013-09-06)
Lysine carbamylation, a post-translational modification, facilitates metal coordination for specific enzymatic activities. We have determined structures of the vertebrate dihydropyrimidinase from Tetraodon nigroviridis (TnDhp) in various states: the apoenzyme as well as two forms of the holoenzyme with one and
E A Lance et al.
Analytical biochemistry, 133(2), 492-501 (1983-09-01)
Potentiometric, visible, infrared, electron spin, and nuclear magnetic resonance studies of the complexation of N-(2-acetamido)iminodiacetic acid (H2ADA) by Ca(II), Mg(II), Mn(II), Zn(II), Co(II), Ni(II), and Cu(II) are reported. Ca(II) and Mg(II) were found not to form 2:1 ADA2- to M(II)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service