30-2630
Tin standard solution
suitable for atomic absorption spectrometry, 1000 ppm Sn, 1 mg/mL Sn
Synonym(s):
Tin(II) chloride solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
SnCl2
CAS Number:
Molecular Weight:
189.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Recommended Products
grade
SAJ special grade
form
liquid
availability
available only in Japan
concentration
1 mg/mL Sn
1000 ppm Sn
suitability
suitable for atomic absorption spectrometry
storage temp.
15-25°C
SMILES string
[Sn]
InChI
1S/Sn
InChI key
ATJFFYVFTNAWJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qaisar Nadeem et al.
Pakistan journal of pharmaceutical sciences, 26(2), 353-357 (2013-03-05)
N-(2-Hydroxybenzyl)-2-amino-2-deoxy-D-glucose (NHADG) was synthesized by conjugation of salicylaldehyde to glucosamine. The obtained compound was well characterized via different analytical techniques. Labeling of the synthesized compound with technetium-99m ((99m)Tc) in pertechnetate form ((99m)Tc O4-) was carried out via chelation reaction in
José Carlos Pelielo De Mattos et al.
Molecules (Basel, Switzerland), 17(11), 12974-12983 (2012-11-03)
Radiopharmaceuticals are employed in patient diagnostics and disease treatments. Concerning the diagnosis aspect, technetium-99m (99mTc) is utilized to label radiopharmaceuticals for single photon computed emission tomography (SPECT) due to its physical and chemical characteristics. 99mTc fixation on pharmaceuticals depends on
Betul Cekic et al.
Acta cirurgica brasileira, 27(9), 606-610 (2012-09-01)
Since Technetium-99m ((99m)Tc) has favorable physical and chemical characteristics, it is widely used radioisotope in Nuclear Medicine. However, stannous dichloride (SnCl(2)) has been widely used as a reducing agent in labeling procedure of pharmaceutical with radionuclide, it has been realized
Aili Sun et al.
Talanta, 88, 259-264 (2012-01-24)
The Zn-Sn nanoparticles/multiwall carbon nanotubes (Zn-SnNPs/MWNTs) nanocomposite film was prepared by electrodeposition of ZnCl(2) and SnCl(2) on MWNTs simultaneously in Ethaline ionic liquids. Then, based on immobilizing hemoglobin (Hb) within a novel Zn-SnNPs/MWNTs nanocomposite film (Hb/Zn-SnNPs/MWNTs), a novel hydrogen peroxide
Matthew Currie et al.
Inorganic chemistry, 52(4), 1710-1721 (2012-06-09)
The anionic speciation of chlorostannate(II) ionic liquids, prepared by mixing 1-alkyl-3-methylimidazolium chloride and tin(II) chloride in various molar ratios, χ(SnCl2), was investigated in both solid and liquid states. The room temperature ionic liquids were investigated by (119)Sn NMR spectroscopy, X-ray
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service