Skip to Content
Merck
All Photos(1)

Key Documents

24-6180

Sigma-Aldrich

1,2-Propanediol

SAJ first grade, ≥99.0%

Synonym(s):

Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥99.0%
bp:
187 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)

grade

SAJ first grade

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.0%

form

viscous liquid

autoignition temp.

779 °F

expl. lim.

12.5 %

availability

available only in Japan

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gai Ling Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(1-2), 49-57 (2002-10-26)
To achieve a therapeutical effect of the anti-Parkinson's drug R-apomorphine via iontophoresis delivery, enhancement strategies in vitro were explored using three structurally related enhancers, lauric acid (LA), dodecyltrimethylammonium bromide (DTAB) and Laureth-3 oxyethylene ether (C(12)EO(3)). Human stratum corneum and shed
R S Rajasenan et al.
Toxicology and applied pharmacology, 135(1), 89-99 (1995-11-01)
Engrafting components of human immune systems in severe combined immunodeficient (SCID) mice has been utilized to investigate the pathogenesis of several human autoimmune diseases and may provide a model for studying idiosyncratic drug toxicity. The purpose of this investigation was
C Fernandez et al.
Journal of pharmaceutical and biomedical analysis, 24(1), 155-165 (2000-12-07)
The aim of this study was to determine the skin penetration of benzophenone-3 in vitro and in vivo in order to investigate a possible influence of formulation. Six different vehicles, three solvents and three different emulsion types were evaluated in
Rong-Yao Wang et al.
Physical chemistry chemical physics : PCCP, 15(9), 3313-3319 (2013-01-31)
Understanding the role of kinetics in fiber network microstructure formation is of considerable importance in engineering gel materials to achieve their optimized performances/functionalities. In this work, we present a new approach for kinetic-structure analysis for fibrous gel materials. In this
J O Funk et al.
Contact dermatitis, 31(4), 236-241 (1994-10-01)
Evaluation of dermatitis associated with propylene glycol application or ingestion remains a challenge. The research dealing with skin reactions to propylene glycol is revisited and new aspects for future research are outlined. Based on literature review and our own observations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service