24-5550
Potassium standard solution
suitable for atomic absorption spectrometry, 1000 ppm K+, 1 mg/mL K+
Synonym(s):
Potassium chloride solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Recommended Products
grade
SAJ special grade
form
liquid
availability
available only in Japan
concentration
1 mg/mL K+
1000 ppm K+
suitability
suitable for atomic absorption spectrometry
SMILES string
[Cl-].[K+]
InChI
1S/ClH.K/h1H;/q;+1/p-1
InChI key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rebecca M Corrigan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 9084-9089 (2013-05-15)
Nucleotide signaling molecules are important messengers in key pathways that allow cellular responses to changing environments. Canonical secondary signaling molecules act through specific receptor proteins by direct binding to alter their activity. Cyclic diadenosine monophosphate (c-di-AMP) is an essential signaling
Francesca Marcon et al.
Biochemical pharmacology, 85(10), 1555-1573 (2013-03-19)
Envenomation by Australian copperheads results mainly in muscle paralysis largely attributed to the presence of postsynaptic α-neurotoxins. However, poorly reversible neurotoxic effects suggest that these venoms may contain snake presynaptic phospholipase A2 neurotoxins (SPANs) that irreversibly inhibit neurotransmitter release. Using
Alec N Salt et al.
The Journal of the Acoustical Society of America, 133(3), 1561-1571 (2013-03-08)
Responses of the ear to low-frequency and infrasonic sounds have not been extensively studied. Understanding how the ear responds to low frequencies is increasingly important as environmental infrasounds are becoming more pervasive from sources such as wind turbines. This study
Yuriko Furukawa et al.
Biochimica et biophysica acta, 1830(9), 4329-4333 (2013-03-05)
Transplantation is one potential clinical application of neural stem cells (NSCs). However, it is very difficult to monitor/control NSCs after transplantation and so provide effective treatment. Electrical measurement using a poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT-PSS) modified microelectrode array (MEA) is a biocompatible, non-invasive
Anusha Priya et al.
Biochimica et biophysica acta, 1833(12), 2745-2756 (2013-07-23)
N-Methyl-d-aspartate (NMDA) receptors are major glutamatergic receptors involved in most excitatory neurotransmission in the brain. The transcriptional regulation of NMDA receptors is not fully understood. Previously, we found that the GluN1 and GluN2B subunits of the NMDA receptor are regulated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service