24-3120
Picric acid solution
Synonym(s):
2,4,6-Trinitrophenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Beilstein:
423400
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Recommended Products
grade
reagent
form
liquid
availability
available only in Japan
density
1.005 g/cm3
SMILES string
Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H
InChI key
OXNIZHLAWKMVMX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Supplementary Hazards
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dustin Staloch et al.
Journal of molecular medicine (Berlin, Germany), 93(10), 1085-1093 (2015-07-05)
The current study aims to identify the pro-fibrogenic role of Gremlin, an endogenous antagonist of bone morphogenetic proteins (BMPs) in chronic pancreatitis (CP). CP is a highly debilitating disease characterized by progressive pancreatic inflammation and fibrosis that ultimately leads to
Debby Kruijsen et al.
Journal of virology, 87(13), 7550-7557 (2013-05-03)
Infants are protected from a severe respiratory syncytial virus (RSV) infection in the first months of life by maternal antibodies or by prophylactically administered neutralizing antibodies. Efforts are under way to produce RSV-specific antibodies with increased neutralizing capacity compared to
D Toki et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(9), 2126-2136 (2014-10-14)
The aim of this study was to investigate the role of infiltrating macrophages in renal allograft fibrosis. Forty-six protocol renal allograft biopsies obtained 1 year after transplantation were stained with Sirius red to quantify fibrosis and double stained with CD68
Giuliana Magri et al.
Nature immunology, 15(4), 354-364 (2014-02-25)
Innate lymphoid cells (ILCs) regulate stromal cells, epithelial cells and cells of the immune system, but their effect on B cells remains unclear. Here we identified RORγt(+) ILCs near the marginal zone (MZ), a splenic compartment that contains innate-like B
Zhonghua Xiang et al.
Macromolecular rapid communications, 33(14), 1184-1190 (2012-04-18)
Three porous luminescent covalent--organic polymers (COPs) have been synthesized through self-polycondensation of the monomers of tris(4-bromophenyl)amine, 1,3,5-tris(4-bromophenyl)benzene, and 2,4,6-tris-(4-bromo-phenyl)-[1,3,5]triazine by using Ni-catalyzed Yamamoto reaction. All the COP materials possess not only high Brunauer-Emmett-Teller (BET) specific surface area of about 2000
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service