Skip to Content
Merck
All Photos(1)

Documents

24-0910

Sigma-Aldrich

Pentane

SAJ first grade, ≥96.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.48 (vs air)

vapor pressure

26.98 psi

Assay

≥96.0%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

availability

available only in Japan

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Maruyama et al.
Bioorganic & medicinal chemistry letters, 20(22), 6661-6666 (2010-10-05)
We have proposed a multi-template approach for drug development, focusing on similar fold structures of proteins, and have effectively generated lead compounds for several drug targets. Modification of these polypharmacological lead compounds is then needed to generate target-selective compounds. In
Michael Polet et al.
Biomedical chromatography : BMC, 27(2), 259-266 (2012-07-21)
The administration of anabolic steroids is one of the most important issues in doping control and is detectable through a change in the carbon isotopic composition of testosterone and/or its metabolites. Gas chromatography-combustion-isotope ratio mass spectrometry (GC-C-IRMS), however, remains a
Dongsheng Guo et al.
Journal of hazardous materials, 186(2-3), 1788-1793 (2011-01-18)
The solvents n-pentane, methylene dichloride, ethyl ether and dodecylbenzenesulphonic acid sodium were used to regenerate exhausted activated carbon used in the process of treating coking wastewater, and the efficiency, ability, and optimum conditions of the different solvents on this regeneration
Fuming Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9174-9181 (2011-07-01)
Graphene oxide (GO) can be viewed as an amphiphilic soft material, which form thin films at organic solvent-water interfaces. However, organic solvent evaporation provides little driving force, which results in slow GO transfer in aqueous phase, thus dawdling GO film
J B Gilman et al.
Environmental science & technology, 47(3), 1297-1305 (2013-01-16)
An extensive set of volatile organic compounds (VOCs) was measured at the Boulder Atmospheric Observatory (BAO) in winter 2011 in order to investigate the composition and influence of VOC emissions from oil and natural gas (O&NG) operations in northeastern Colorado.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service