Skip to Content
Merck
All Photos(1)

Documents

19-4150

Sigma-Aldrich

Methyl salicylate

SAJ first grade, ≥98.0%

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

847 °F

availability

available only in Japan

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taraka R Donti et al.
Disease models & mechanisms, 7(2), 271-280 (2013-11-26)
Mutations in subunits of succinyl-CoA synthetase/ligase (SCS), a component of the citric acid cycle, are associated with mitochondrial encephalomyopathy, elevation of methylmalonic acid (MMA), and mitochondrial DNA (mtDNA) depletion. A FACS-based retroviral-mediated gene trap mutagenesis screen in mouse embryonic stem
Agata Bartyzel et al.
Analytica chimica acta, 707(1-2), 204-209 (2011-10-27)
N,N'-bis-(α-methylsalicylidene)-2,2-dimethyl-1,3-propanediimine (SBTD) modified silica gel was prepared and used as sorbent for solid phase extraction of Cr(III) ions from aqueous solution. This sorbent showed a high sorption affinity for Cr(III) while recovery of Cr(VI) was very low. The analyte ion
Michiel van Wijk et al.
PloS one, 6(7), e21742-e21742 (2011-07-19)
Insect herbivory induces plant odors that attract herbivores' natural enemies. Assuming this attraction emerges from individual compounds, genetic control over odor emission of crops may provide a rationale for manipulating the distribution of predators used for pest control. However, studies
Nadir Erbilgin et al.
Tree physiology, 32(8), 946-957 (2012-06-05)
Coniferous trees have both constitutive and inducible defences that deter or kill herbivores and pathogens. We investigated constitutive and induced monoterpene responses of jack pine (Pinus banksiana Lamb.) to a number of damage types: a fungal associate of the mountain
Richard Radnovich et al.
Journal of pain research, 7, 727-735 (2014-12-20)
Treatment for pain due to shoulder impingement syndrome (SIS) typically begins conservatively with nonsteroidal anti-inflammatory drugs and physical therapy and can include subacromial injection of corticosteroids, particularly in patients unresponsive to conservative measures. The heated lidocaine/tetracaine (HLT) patch has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service