Skip to Content
Merck
All Photos(1)

Documents

19-4120

Sigma-Aldrich

Methyl Red

JIS special grade

Synonym(s):

2-(4-Dimethylaminophenylazo)benzoic acid, 4-Dimethylaminoazobenzene-2′-carboxylic acid, Acid Red 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
269.30
Colour Index Number:
13020
Beilstein:
750102
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

mp

179-182 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)/b17-16+

InChI key

CEQFOVLGLXCDCX-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chan-Ju Wang et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 66(Pt 1), 2-7 (2010-01-09)
Azoreductase 1 from Pseudomonas aeruginosa strain PAO1 (paAzoR1) catalyses the activation of the prodrug balsalazide and reduces the azo dye methyl red using reduced nicotinamide adenine dinucleotide cofactor as an electron donor. To investigate the mechanism of the enzyme, a
Oana Alexandru et al.
Journal of neuro-oncology, 102(1), 9-18 (2010-07-17)
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of
X D Zhang et al.
Journal of hazardous materials, 170(2-3), 883-887 (2009-06-09)
An aqueous C.I. Acid Red 2 solution was decolorized by electrolysis using iron as anode. The decolorization mechanism was investigated through experimental observations on the electrochemical behavior of C.I. Acid Red 2 on Pt rotating disk electrode, UV-visible spectra of
Taiga Fujii et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(35), 10865-10872 (2012-07-26)
Asymmetric dye clusters with a single fluorophore (Cy3) and multiple quenchers (4'-methylthioazobenzene-4-carboxylate, methyl red, and 4'-dimethylamino-2-nitroazobenzene-4-carboxylate) were prepared. The dye and one-to-five quenchers were tethered through D-threoninol to opposite strands of a DNA duplex. NMR analysis revealed that the clusters
Danmeng Shuai et al.
Environmental science & technology, 44(5), 1773-1779 (2010-02-11)
Azo dyes are widespread pollutants and potential cocontaminants for nitrate; we evaluated their effect on catalytic reduction of a suite of oxyanions, diatrizoate, and N-nitrosodimethylamine (NDMA). The azo dye methyl orange significantly enhanced (less than or equal to a factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service